Merck
  • Detection of pancreatic cancer with normal carbohydrate antigen 19-9 using protein chip technology.
Detection of pancreatic cancer with normal carbohydrate antigen 19-9 using protein chip technology.
World journal of gastroenterology (2014-10-31)
Xiao-Li Jin, Bin Xu, Yu-Lian Wu
ZUSAMMENFASSUNG
To develop a method to differentiate pancreatic cancer patients from healthy or benign individuals when carbohydrate antigen (CA) 19-9 is normal. Forty-one serum samples from patients with pancreatic lesions and blood samples from 20 healthy individuals were collected at the first stage of the experiment according to the enrolment criteria. General characteristics and some clinical features were carefully compared to ensure that the results were reasonable. All the blood samples were analyzed by surface-enhanced laser desorption/ionization time-of-flight mass spectrometry (SELDI-TOF-MS) combined with CM10 chips and a related bioinformatics analysis program to generate diagnostic models with different proteins. Forty-seven consecutive samples were tested at the next stage to verify the veracity and efficiency of the models. The sex, age, and serum CA19-9 levels among the three groups (malignant, benign, and healthy) were statistically matched (P values were 0.957, 0.145, and 0.382, respectively). Two patterns were generated. Pattern 1 with four proteins theoretically had a specificity and sensitivity of 100% in distinguishing pancreatic cancer from healthy individuals, while it was 86.7% and 86.4%, respectively, in the subsequent practical verification. The positive predictive value (PPV) of the model was 86.4%. One of the four proteins was expressed highly in pancreatic cancer while the other three were expressed weakly. Pattern 2 consisted of six proteins that showed a specificity of 70.0% and sensitivity of 77.3% for differentiating malignancy from benign tumors. Its PPV reached 85.0%. Only one of these six proteins showed high expression in the malignant group. SELDI-TOF-MS may facilitate diagnosis or differential diagnosis of pancreatic cancer when CA19-9 is normal. Pattern 1 may serve as a useful screening tool.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Sinapinsäure, ≥98%, powder
Supelco
Sinapinsäure, suitable for matrix substance for MALDI-MS, ≥99.0% (T)
Supelco
Sinapinsäure, suitable for matrix substance for MALDI-MS, ≥99.5%, Ultra pure
Supelco
trans-Sinapinsäure, analytical standard