Merck
  • Neuroprotective effects of Z-ajoene, an organosulfur compound derived from oil-macerated garlic, in the gerbil hippocampal CA1 region after transient forebrain ischemia.
Neuroprotective effects of Z-ajoene, an organosulfur compound derived from oil-macerated garlic, in the gerbil hippocampal CA1 region after transient forebrain ischemia.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association (2014-07-06)
Dae Young Yoo, Woosuk Kim, Sung Min Nam, Miyoung Yoo, Sanghee Lee, Yeo Sung Yoon, Moo-Ho Won, In Koo Hwang, Jung Hoon Choi
ZUSAMMENFASSUNG
The neuroprotective effects of two isomers (Z- and E-) of ajoene, a major compound in oil-macerated garlic products, against ischemic damage were investigated in the gerbil hippocampus. Vehicle (corn oil), Z- or E-ajoenes (25 mg/kg) was orally administered 30 min prior to the induction of transient forebrain ischemia by occlusion of the common carotid arteries for 5 min. One day after ischemia/reperfusion (I/R), I/R-induced hyperactivity significantly reduced in the E- and Z-ajoene-treated groups, compared to that in the vehicle-treated group 5 days after I/R, the number of cresyl violet-positive neurons in the E- and Z-ajoene-treated groups increased, compared to that in the vehicle-treated group. Reactive gliosis in the CA1 region of E- and Z-ajoene-treated groups reduced, compared to that in the vehicle-treated group. These neuroprotective effects were more prominent in animals treated with Z-ajoene, than in those treated with E-ajoene. In addition, Z-ajoene significantly decreased lipid peroxidation, as indicated by 4-hydroxy-2-nonenal levels in hippocampal homogenates, compared to that observed in the vehicle-treated group at a range of time points after I/R. These results suggested that Z-ajoene protected against I/R-induced delayed neuronal death and gliosis by reducing lipid peroxidation in the gerbil hippocampal CA1 region.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Essigsäure, glacial, ACS reagent, ≥99.7%
Sigma-Aldrich
Essigsäure, glacial, ReagentPlus®, ≥99%
Sigma-Aldrich
Essigsäure, glacial, ≥99.99% trace metals basis
Sigma-Aldrich
Essigsäure -Lösung, suitable for HPLC
Sigma-Aldrich
3,3′-Diaminobenzidin -tetrahydrochlorid, tablet, 10 mg substrate per tablet
Sigma-Aldrich
Essigsäure, suitable for luminescence, BioUltra, ≥99.5% (GC)
USP
Eisessig, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
5α-Androstan-17β-ol-3-on, ≥97.5%
Supelco
Essigsäure, analytical standard
Sigma-Aldrich
Essigsäure, ≥99.5%, FCC, FG
Sigma-Aldrich
Essigsäure, natural, ≥99.5%, FG
Sigma-Aldrich
5α-Androstan-17β-ol-3-on, purum, ≥99.0% (TLC)
Supelco
5α-Androstan-17β-ol-3-on, VETRANAL®, analytical standard
Sigma-Aldrich
Essigsäure-12C2, 99.9 atom % 12C
Millipore
Bifido Selektives Supplement B, suitable for microbiology