Merck
  • Perfluorodecalin-soluble fluorescent dyes for the monitoring of circulating nanocapsules with intravital fluorescence microscopy.
Perfluorodecalin-soluble fluorescent dyes for the monitoring of circulating nanocapsules with intravital fluorescence microscopy.
Journal of microencapsulation (2014-06-26)
J Laudien, D Naglav, C Groβ-Heitfeld, K B Ferenz, H de Groot, C Mayer, S Schulz, A Schnepf, M Kirsch
ZUSAMMENFASSUNG
Perfluorodecalin (PFD) is an established artificial oxygen carrier due to its physical capability to solve the respiratory gases oxygen and carbon dioxide. PFD-filled poly(n-butyl-cyanoacrylate) (PACA) nanocapsules are already discussed as effective artificial oxygen carriers, and their principal suitability for intravenous administration had been shown. To further elucidate their action in vivo, it is imperative to characterise their preclinical safety and particularly their biodistribution. For these purposes, intravital fluorescence microscopy would display an attractive technique in order to monitor the PACA nanocapsules in vivo, but unfortunately, it is impossible to stain the PACA nanocapsules with a fluorescent dye fulfilling special criteria required for in vivo microscopy. In order to develop such a dye, a long-chained fluorinated thiol was used to modify a BODIPY derivative that is a highly fluorescent organic compound belonging to the difluoro-boraindacene family, as well as to functionalise mesoscopic systems, such as CdSe/ZnS-quantum dots and gold nanoparticles. Furthermore, a functionalisation of porphyrin derivatives was investigated by placing divalent ions in the centre of these systems. Due to the high solubility of all synthesised dyes in PFD, it should be possible to stain PFD-filled particles in general. However, only the functionalised BODIPY derivative was suitable for in vivo monitoring of the PFD-filled PACA nanocapsules.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Heptan, suitable for HPLC, ≥99%
Sigma-Aldrich
Heptan, ReagentPlus®, 99%
Sigma-Aldrich
Heptan, HPLC Plus, for HPLC, GC, and residue analysis, 99%
Sigma-Aldrich
Heptan, anhydrous, 99%
Sigma-Aldrich
Heptan, suitable for HPLC, ≥96%
Supelco
Heptan, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Heptan, analytical standard
Sigma-Aldrich
Heptan, biotech. grade, ≥99%