Merck
  • Development of serous ovarian cancer is associated with the expression of homologous recombination pathway proteins.
Development of serous ovarian cancer is associated with the expression of homologous recombination pathway proteins.
Pathology oncology research : POR (2014-04-23)
Qingqing Ye, Li Chen, Xiaolu Yin, Yuan Jie Charles Liu, Qunsheng Ji, Enfeng Zhao
ZUSAMMENFASSUNG
To investigate the expressions of key markers in the homologous recombination (HR) pathway and the correlation with clinicopathological parameters in serous ovarian cancer (SOC). We analyzed the protein expression of MRE11, MDC1, ATM, ATR and BRCA1 by immunohistochemistry (IHC) in 97 SOC samples, and correlated with clinical parameters including age, tumor grades, clinical stage, status of menstruation and chemotherapy. Low expression of MRE11 and MDC1 was detected in 14.4 % and 3.1 % of the patient samples, and negative expression of ATM, ATR and BRCA1 was found in 11.3 %, 6.3 % and 29.9 % of the patient samples, respectively. ATR deficiency was significantly associated with menopause (P = 0.025), strong expression of ATM (P = 0.017) and MRE11 (P = 0.040) with pre-menopausal SOC, strong expression of MRE11 (P = 0.016) with low tumor grade, and strong expression of BRCA1 (P = 0.015) with early clinical stage. In addition, low expression of MRE11 was strongly associated with negativity of ATR (P < 0.001) and BRCA1 (P = 0.004) Furthermore, ATR deficiency was associated with low expression of ATM (P = 0.028) and loss expression of BRCA1 (P = 0.009). Our results suggest that reduced expression or loss of proteins in HR pathway is associated with SOC development. Abnormality of MRE11 and BRCA1 are strongly associated with late clinical stage in SOC patients.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Ethyl alcohol, Pure 200 proof, Molecular Biology
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure 190 proof, for molecular biology
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Ethanol, BioUltra, Molecular Biology, ≥99.8%, (absolute alcohol, without additive, A15 o1)
Supelco
Ethanol, wasserfrei, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ethanol, purum, fine spirit, denaturated with 4.8% methanol, F25 METHYL1, ~96% (based on denaturant-free substance)
Supelco
Ethanol, standard for GC
Sigma-Aldrich
Ethanol
Sigma-Aldrich
Ethanol, tested according to Ph. Eur.
Supelco
Ethanol -Lösung, certified reference material, 2000 μg/mL in methanol
USP
Dehydrated Alcohol, United States Pharmacopeia (USP) Reference Standard