Merck
  • Global metabolite profiling of synovial fluid for the specific diagnosis of rheumatoid arthritis from other inflammatory arthritis.
Global metabolite profiling of synovial fluid for the specific diagnosis of rheumatoid arthritis from other inflammatory arthritis.
PloS one (2014-06-03)
Sooah Kim, Jiwon Hwang, Jinhua Xuan, Young Hoon Jung, Hoon-Suk Cha, Kyoung Heon Kim
ZUSAMMENFASSUNG
Currently, reliable biomarkers that can be used to distinguish rheumatoid arthritis (RA) from other inflammatory diseases are unavailable. To find possible distinctive metabolic patterns and biomarker candidates for RA, we performed global metabolite profiling of synovial fluid samples. Synovial fluid samples from 38 patients with RA, ankylosing spondylitis, Behçet's disease, and gout were analyzed by gas chromatography/time-of-flight mass spectrometry (GC/TOF MS). Orthogonal partial least-squares discriminant and hierarchical clustering analyses were performed for the discrimination of RA and non-RA groups. Variable importance for projection values were determined, and the Wilcoxon-Mann-Whitney test and the breakdown and one-way analysis of variance were conducted to identify potential biomarkers for RA. A total of 105 metabolites were identified from synovial fluid samples. The score plot of orthogonal partial least squares discriminant analysis showed significant discrimination between the RA and non-RA groups. The 20 metabolites, including citrulline, succinate, glutamine, octadecanol, isopalmitic acid, and glycerol, were identified as potential biomarkers for RA. These metabolites were found to be associated with the urea and TCA cycles as well as fatty acid and amino acid metabolism. The metabolomic analysis results demonstrated that global metabolite profiling by GC/TOF MS might be a useful tool for the effective diagnosis and further understanding of RA.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Pyridin, ACS reagent, ≥99.0%
Sigma-Aldrich
Pyridin, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Pyridin, anhydrous, 99.8%
Sigma-Aldrich
Pyridin, ≥99%
Sigma-Aldrich
Pyridin, biotech. grade, ≥99.9%
Supelco
Pyridin, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Pyridin, analytical standard