Merck
Induction of vascular leak syndrome by tumor necrosis factor-alpha alone.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie (2015-03-18)
Kyung-Yeon Park, Sung-Jo Kim, Euichaul Oh, Tae-Hwe Heo
ZUSAMMENFASSUNG
Although TNF-α possesses promising anticancer activity, clinical application is limited partly due to cardiovascular toxicities. TNF-α effects on vessels are likely related to vascular toxicity, but much remains poorly understood. Similarly, IL-2 is an attractive treatment option for cancers but its clinical use is limited by the side effect of vascular leak syndrome (VLS). We report here that TNF-α alone can trigger VLS. Administration of recombinant TNF-α induced VLS in normal mice and TNF-α transgenic mice exhibited VLS. Perivascular infiltrates in the lungs and specific cytokines in serum were observed in VLS-induced mice. This study shed a new light on the critical role of the TNF-α in IL-2-induced or non IL-2-induced VLS and provides important points to TNF-α therapy.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Formamid, ReagentPlus®, ≥99.0% (GC)
Sigma-Aldrich
Formamid, ≥99.5% (GC), BioReagent, Molecular Biology
Sigma-Aldrich
Formamid, BioUltra, Molecular Biology, ≥99.5% (T)
Sigma-Aldrich
Formamid, spectrophotometric grade, ≥99%
Supelco
Formamid -Lösung, suitable for NMR (reference standard), 90% in DMSO-d6 (99.9 atom % D), NMR tube size 5 mm × 8 in.
Sigma-Aldrich
Formamid -Lösung, suitable for NMR (reference standard), 90% in DMSO-d6 (99.9 atom % D), NMR tube size 10 mm × 8 in.
Supelco
Formamid -Lösung, suitable for NMR (reference standard), 90% in DMSO-d6 (99.9 atom % D), NMR tube size 5 mm × 8 in.