Merck
  • Selection of indigenous lactic acid bacteria to reinforce the intestinal microbiota of newly hatched chicken: relevance of in vitro and ex vivo methods for strains characterization.
Selection of indigenous lactic acid bacteria to reinforce the intestinal microbiota of newly hatched chicken: relevance of in vitro and ex vivo methods for strains characterization.
Research in veterinary science (2014-07-01)
Jaime D Babot, Eloy Argañaraz-Martínez, Lucila Saavedra, María C Apella, Adriana Perez Chaia
ZUSAMMENFASSUNG
Based on the natural benefits of the indigenous microbiota, lactic acid bacteria (LAB) from poultry origin were isolated from hens and broilers intestine, and their probiotic potential was further studied. The tolerance to digestion, adhesion, capture of a mannose-binding lectin, absence of virulent factors and antibiotic resistances were studied. Different in vitro and ex vivo assays were performed to select tolerant and adherent strains because standardized protocols have not been defined. Fourteen strains highly tolerant to gastrointestinal digestion were genetically identified. Hydrophobic surfaces were not required for the bacterial adhesion and only nine strains adhered ex vivo to the intestinal mucosa. Three strains captured a lectin of the same specificity of Type-1 fimbriae. Virulence factors were absent but some strains evidenced multiple antibiotic resistances. These results provide bases for a future standardization of methods for the selection of probiotic strains intended to reinforce the microbiota of newly hatched chickens.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Milchsäure, meets USP testing specifications
Sigma-Aldrich
Milchsäure, 88%, FCC
Sigma-Aldrich
Milchsäure, natural, ≥85%
Sigma-Aldrich
Milchsäure -Lösung, ACS reagent, ≥85%
Sigma-Aldrich
DL-Milchsäure, ~90% (T)
Supelco
Milchsäure, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Milchsäure, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
DL-Milchsäure, 85 % (w/w), syrup