Merck
Overexpression of MAGE-A9 predicts unfavorable outcome in breast cancer.
Experimental and molecular pathology (2014-12-03)
Xin Xu, Xiaojun Tang, Meiping Lu, Qi Tang, Huilin Zhang, Huijun Zhu, Ning Xu, Dawei Zhang, Lin Xiong, Yuan Mao, Jin Zhu
ZUSAMMENFASSUNG
Melanoma-associated antigens (MAGEs) are a group of well-characterized members of the cancer/testis antigen (CTA) family, which is one of the largest groups of human tumor-associated antigens. MAGE-A9 is a particular member in the context of the MAGE-A gene family and was defined as presenting prognostic relevance in certain type of human cancer. However, the expression of MAGE-A9 in invasive ductal breast cancer (IDC) and the relationship with the clinical attributes of IDC, especially prognostic characteristic, remain poorly understood. In this present study, one-step quantitative reverse transcription polymerase chain reaction (18 fresh-frozen IDC tissues and corresponding non-cancerous tissues) and immunohistochemistry by tissue microarrays (82 IDC tissue samples and 70 matched tumor-adjacent non-cancerous tissue samples) were performed to characterize expression of the MAGE-A9 gene in IDC. Kaplan-Meier survival and Cox regression analyses were performed to evaluate the prognosis of IDC. The results of qPCR and IHC analysis showed that the expression of MAGE-A9 in IDC was significantly higher than that in corresponding non-cancerous tissue. Moreover, the expression level of MAGE-A9 protein in IDC was significantly related to histological grade (p = 0.011) and distant metastasis (p = 0.019). Multivariate analysis with the Cox regression model showed that MAGE-A9 protein expression (p = 0.009), histological grade (p = 0.014), lymph node metastasis (p = 0.012) and distant metastasis (p = 0.011) were independent prognostic factors for overall survival of IDC patients. The data suggest that MAGE-A9 expression is correlated with malignant attributes of IDC and it may serve as a novel prognostic factor and an ideal candidate for targeted therapy in IDC treatment.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Trimesinsäure, 95%
Sigma-Aldrich
Trimethylaluminium -Lösung, 2.0 M in toluene
Sigma-Aldrich
Trimethylaluminium, 97%
Sigma-Aldrich
Trimethylaluminium, packaged for use in deposition systems
Sigma-Aldrich
Trimethylaluminium -Lösung, 2.0 M in hexanes
Sigma-Aldrich
Tris(tert-butoxy)silanol, 99.999%
Sigma-Aldrich
Trimethylaluminium -Lösung, 2.0 M in heptane