Merck
MicroRNA-145 targets MUC13 and suppresses growth and invasion of pancreatic cancer.
Oncotarget (2014-10-04)
Sheema Khan, Mara C Ebeling, Mohd S Zaman, Mohammed Sikander, Murali M Yallapu, Neeraj Chauhan, Ashley M Yacoubian, Stephen W Behrman, Nadeem Zafar, Deepak Kumar, Paul A Thompson, Meena Jaggi, Subhash C Chauhan
ZUSAMMENFASSUNG
Pancreatic cancer has a poor prognosis due to late diagnosis and ineffective therapeutic multimodality. MUC13, a transmembrane mucin is highly involved in pancreatic cancer progression. Thus, understanding its regulatory molecular mechanisms may offer new avenue of therapy for prevention/treatment of pancreatic cancer. Herein, we report a novel microRNA (miR-145)-mediated mechanism regulating aberrant MUC13 expression in pancreatic cancer. We report that miR-145 expression inversely correlates with MUC13 expression in pancreatic cancer cells and human tumor tissues. miR-145 is predominantly present in normal pancreatic tissues and early Pancreatic Ductal Adenocarcinoma (PDAC) precursor lesions (PanIN I) and is progressively suppressed over the course of development from PanIN II/III to late stage poorly differentiated PDAC. We demonstrate that miR-145 targets 3' untranslated region of MUC13 and thus downregulates MUC13 protein expression in cells. Interestingly, transfection of miR-145 inhibits cell proliferation, invasion and enhances gemcitabine sensitivity. It causes reduction of HER2, P-AKT, PAK1 and an increase in p53. Similar results were found when MUC13 was specifically inhibited by shRNA directed at MUC13. Additionally, intratumoral injections of miR-145 in xenograft mice inhibited tumor growth via suppression of MUC13 and its downstream target, HER2. These results suggest miR-145 as a novel regulator of MUC13 in pancreatic cancer.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Propidiumjodid, ≥94.0% (HPLC)
Sigma-Aldrich
Nitrotetrazolium-Blauchlorid, ≥90.0% (HPLC)
Sigma-Aldrich
Propidiumjodid -Lösung
Sigma-Aldrich
Propidiumjodid, ≥94% (HPLC)
Sigma-Aldrich
Nitrotetrazolium-Blauchlorid, powder, electrophoresis grade
Sigma-Aldrich
MISSION® esiRNA, targeting human MUC13