Merck
  • Synthesis and biological evaluation of imidazo[1,5-a]pyridine-benzimidazole hybrids as inhibitors of both tubulin polymerization and PI3K/Akt pathway.
Synthesis and biological evaluation of imidazo[1,5-a]pyridine-benzimidazole hybrids as inhibitors of both tubulin polymerization and PI3K/Akt pathway.
Organic & biomolecular chemistry (2014-10-31)
Ahmed Kamal, A V Subba Rao, V Lakshma Nayak, N V Subba Reddy, Konderu Swapna, G Ramakrishna, Mallika Alvala
ZUSAMMENFASSUNG
A series of imidazo[1,5-a]pyridine-benzimidazole hybrids (5a–aa) were prepared and evaluated for their cytotoxic activity against a panel of sixty human tumor cell lines. Among them compounds 5d and 5l showed significant cytotoxic activity with GI50 values ranging from 1.06 to 14.9 μM and 0.43 to 7.73 μM, respectively. Flow cytometric analysis revealed that these compounds arrest the cell cycle at G2/M phase and induced cell death by apoptosis. The tubulin polymerization assay (IC50 of 5d is 3.25 μM and 5l is 1.71 μM) and immunofluorescence analysis showed that these compounds effectively inhibited the microtubule assembly in human breast cancer cells (MCF-7). Further, the apoptotic effects of compounds were confirmed by Hoechst staining, mitochondrial membrane potential, cytochrome c release, ROS generation, caspase 9 activation and DNA fragmentation analysis. After treatment with these compounds for 48 h, p-PTEN and p-AKT levels were markedly decreased. Moreover, these compounds did not significantly inhibit the normal human embryonic kidney cells, HEK-293. The molecular docking simulations predicted the binding interactions of 5d and 5l with colchicine binding site of the tubulin, which is in compliance with the antiproliferative activity data.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Ethyl alcohol, Pure 200 proof, Molecular Biology
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure 190 proof, for molecular biology
Sigma-Aldrich
Propidiumjodid, ≥94.0% (HPLC)
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Ethanol, BioUltra, Molecular Biology, ≥99.8%, (absolute alcohol, without additive, A15 o1)
Supelco
Ethanol, wasserfrei, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ethanol, purum, fine spirit, denaturated with 4.8% methanol, F25 METHYL1, ~96% (based on denaturant-free substance)
Supelco
Ethanol, standard for GC
Sigma-Aldrich
Propidiumjodid -Lösung
Sigma-Aldrich
2′,7′-Dichlorfluoreszeindiacetat, BioReagent, suitable for fluorescence, ≥95% (HPLC)
Sigma-Aldrich
Propidiumjodid, ≥94% (HPLC)
Sigma-Aldrich
Ethanol
Sigma-Aldrich
Ethanol, tested according to Ph. Eur.
Supelco
Ethanol -Lösung, certified reference material, 2000 μg/mL in methanol
USP
Dehydrated Alcohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
1,3-Propylene sulfite, 99%