Merck
  • A common polymorphism in extracellular superoxide dismutase affects cardiopulmonary disease risk by altering protein distribution.
A common polymorphism in extracellular superoxide dismutase affects cardiopulmonary disease risk by altering protein distribution.
Circulation. Cardiovascular genetics (2014-08-03)
John M Hartney, Timothy Stidham, David A Goldstrohm, Rebecca E Oberley-Deegan, Michael R Weaver, Zuzana Valnickova-Hansen, Carsten Scavenius, Richard K P Benninger, Katelyn F Leahy, Richard Johnson, Fabienne Gally, Beata Kosmider, Angela K Zimmermann, Jan J Enghild, Eva Nozik-Grayck, Russell P Bowler
ZUSAMMENFASSUNG
The enzyme extracellular superoxide dismutase (EC-SOD; SOD3) is a major antioxidant defense in lung and vasculature. A nonsynonomous single-nucleotide polymorphism in EC-SOD (rs1799895) leads to an arginine to glycine amino acid substitution at position 213 (R213G) in the heparin-binding domain. In recent human genetic association studies, this single-nucleotide polymorphism attenuates the risk of lung disease, yet paradoxically increases the risk of cardiovascular disease. Capitalizing on the complete sequence homology between human and mouse in the heparin-binding domain, we created an analogous R213G single-nucleotide polymorphism knockin mouse. The R213G single-nucleotide polymorphism did not change enzyme activity, but shifted the distribution of EC-SOD from lung and vascular tissue to extracellular fluid (eg, bronchoalveolar lavage fluid and plasma). This shift reduces susceptibility to lung disease (lipopolysaccharide-induced lung injury) and increases susceptibility to cardiopulmonary disease (chronic hypoxic pulmonary hypertension). We conclude that EC-SOD provides optimal protection when localized to the compartment subjected to extracellular oxidative stress: thus, the redistribution of EC-SOD from the lung and pulmonary circulation to the extracellular fluids is beneficial in alveolar lung disease but detrimental in pulmonary vascular disease. These findings account for the discrepant risk associated with R213G in humans with lung diseases compared with cardiovascular diseases.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Glycin, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Glycin, suitable for electrophoresis, ≥99%
Sigma-Aldrich
Glycin, BioUltra, Molecular Biology, ≥99.0% (NT)
Sigma-Aldrich
Glycin, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, ≥98.5%
Sigma-Aldrich
SOD Bestimmungs-Kit, sufficient for 500 tests
SAFC
Glycin
Supelco
Glycin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Glycin, BioXtra, ≥99% (titration)
Sigma-Aldrich
Glycin, ACS reagent, ≥98.5%
Sigma-Aldrich
Glycin, 99%, FCC
Sigma-Aldrich
Glycin, meets analytical specification of Ph. Eur., BP, USP, 99-101% (based on anhydrous substance)
USP
Glycin, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Glycin, puriss. p.a., reag. Ph. Eur., buffer substance, 99.7-101% (calc. to the dried substance)
Supelco
Glycin, analytical standard, for nitrogen determination according to Kjeldahl method
Glycin, European Pharmacopoeia (EP) Reference Standard
Supelco
Glycin, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Glycin, tested according to Ph. Eur.