Merck
  • Phenotype modifications of T-cells and their shift toward a Th2 response in patients with systemic lupus erythematosus supplemented with different monthly regimens of vitamin D.
Phenotype modifications of T-cells and their shift toward a Th2 response in patients with systemic lupus erythematosus supplemented with different monthly regimens of vitamin D.
Lupus (2015-03-25)
S Piantoni, L Andreoli, M Scarsi, A Zanola, F Dall'Ara, C Pizzorni, M Cutolo, P Airò, A Tincani
ZUSAMMENFASSUNG
Vitamin D receptor is constitutively expressed on the lymphocyte surface. Recent studies highlight that vitamin D may exert actions on T-cells, inhibiting Th1 and Th17 response and enhancing Th2 and T-regulatory (T-reg) function. Thirty-four patients with systemic lupus erythematosus (SLE) were randomly enrolled in a two-year prospective study. In the first year, 16 patients were supplemented with an intensive regimen of cholecalciferol (IR) (300.000 UI of cholecalciferol at baseline and 50.000 UI/monthly as maintenance, 850.000 UI annually), whereas 18 with a standard regimen (SR) (25.000 UI of cholecalciferol monthly, 300.000 UI annually). During the second year, patients were switched to the other arm of treatment. Phenotypic analysis of peripheral T lymphocyte and the quantification of cytokine production from peripheral blood mononuclear cells (PBMCs) were evaluated by flow cytometry. At baseline, no significant difference between the two groups emerged among main T-cell subtypes. Over two years of treatment, we saw an increase in the number of T-reg cells, in the total amount of CD4+CD45RA+CCR7- T-cells, whereas a significant reduction of CD8+CD28- T-cells was observed. In addition, the analysis of PBMCs from eight patients following the IR showed the reduction of the IFN-γ/IL-4 ratio (p = 0.01) among CD8+ T-cells after 12 months. After a long-term of monthly treatment with vitamin D in SLE patients, an enhancement of T-reg cells and the production of Th2 cytokines should be expected.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Phorbol-12-myristat-13-acetat, ≥99% (TLC), film or powder
Sigma-Aldrich
Myristinsäure, Sigma Grade, ≥99%
Sigma-Aldrich
Phorbol-12-myristat-13-acetat, synthetic, ≥98.0% (TLC)
Sigma-Aldrich
Myristinsäure, natural, ≥98.5%, FG
Sigma-Aldrich
Myristinsäure, ≥95%, FCC, FG
Sigma-Aldrich
Myristinsäure, ≥98.0% (GC)
Supelco
Myristinsäure, analytical standard
Supelco
Myristinsäure, Pharmaceutical Secondary Standard; Certified Reference Material