Merck
Detection of formestane abuse by mass spectrometric techniques.
Drug testing and analysis (2014-12-18)
Xavier de la Torre, Cristiana Colamonici, Davide Curcio, Daniel Jardines, Francesco Molaioni, Maria Kristina Parr, Francesco Botrè
ZUSAMMENFASSUNG
Formestane (4-hydroxy-androstenedione) is an aromatase inhibitor prohibited in sports and included, since 2004, in the list of prohibited substances updated yearly by the World Anti-Doping Agency (WADA). Since the endogenous production of formestane has been described, it is mandatory for the anti-doping laboratories to use isotope ratio mass spectrometry (IRMS) to establish the exogenous origin before issuing an adverse analytical finding. The described IRMS methods for formestane detection are time-consuming, requiring usually two consecutive liquid chromatographic sample purifications in order to have final extracts of adequate purity before the mass spectrometric analysis. After establishing a procedure for the determination of the origin of formestane by IRMS without the need of derivatization, and integrated in the overall analytical strategy of the laboratory for pseudo-endogenous steroids, a mass spectrometric analysis by gas chromatography-mass spectrometry (GC-MS) and gas chromatography-tandem mass spectrometry (GC-MS/MS) of formestane metabolites was carried out in order to investigate whether other biomarkers of formestane abuse could be integrated in order to avoid time-consuming and expensive IRMS confirmations for formestane. From the metabolic studies performed, the inclusion of 3β,4α-dihydroxy-5α-androstan-17-one (4α-hydroxy-epiandosterone) in the routine GC-MS procedures has demonstrated to be diagnostic in order to reduce the number of unnecessary confirmations of the endogenous origin of formestane.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Formestan, solid