Merck
Type I collagen prevents amyloid aggregation of hen egg white lysozyme.
Biochemical and biophysical research communications (2014-05-08)
Kriti Dubey, Karunakar Kar
ZUSAMMENFASSUNG
Both collagen and amyloidogenic proteins have an inherent ability to undergo a self-assembly process leading to formation of supramolecular structures. Though our understanding of collagen-amyloid link is very poor, a few experimental evidences have indicated the protective nature of collagen against amyloid fibril formation. To further our understanding of collagen-amyloid relationship, we have explored the role of type I collagen on amyloid-aggregation of lysozyme. Thioflavin-T assay data indicated strong inhibition of both spontaneous and seeded aggregation of lysozyme by collagen. Both chemical and thermal denaturation experiments have showed increased lysozyme stability in the presence of collagen. However, the presence of collagen did not alter lysozyme activity. These findings confirm that type I collagen is capable of blocking or interfering with the amyloid aggregation of lysozyme, and the results may have significant implications for the design of collagen based therapeutics against aggregation of disease linked amyloidogenic proteins.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Glycin, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Glycin, suitable for electrophoresis, ≥99%
Sigma-Aldrich
Glycin, BioUltra, Molecular Biology, ≥99.0% (NT)
Sigma-Aldrich
Glycin, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, ≥98.5%
SAFC
Glycin
Supelco
Glycin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Glycin, BioXtra, ≥99% (titration)
Sigma-Aldrich
Glycin, ACS reagent, ≥98.5%
Sigma-Aldrich
Glycin, 99%, FCC
USP
Glycin, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Glycin, meets analytical specification of Ph. Eur., BP, USP, 99-101% (based on anhydrous substance)
Sigma-Aldrich
Glycin, puriss. p.a., reag. Ph. Eur., buffer substance, 99.7-101% (calc. to the dried substance)
Supelco
Glycin, analytical standard, for nitrogen determination according to Kjeldahl method
Glycin, European Pharmacopoeia (EP) Reference Standard
Supelco
Glycin, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Glycin, tested according to Ph. Eur.