Merck
  • Brain-specific delivery of dopamine mediated by n,n-dimethyl amino group for the treatment of Parkinson's disease.
Brain-specific delivery of dopamine mediated by n,n-dimethyl amino group for the treatment of Parkinson's disease.
Molecular pharmaceutics (2014-07-30)
Yanping Li, Yangyang Zhou, Bowen Qi, Tao Gong, Xun Sun, Yao Fu, Zhirong Zhang
ZUSAMMENFASSUNG
Parkinson's disease (PD) has become one of the most deadly diseases due to a lack of effective treatment. Herein, N-3,4-bis(pivaloyloxy)dopamine-3-(dimethylamino)propanamide (PDDP), a brain-specific derivative of dopamine, was designed and synthesized, which consists of a brain targeted ligand, N,N-dimethyl amino group, and two dipivaloyloxy groups for lipophilic modification. PDDP was investigated both in vitro and in vivo by comparing with L-DOPA and another derivative (BPD) without N,N-dimethyl amino group. PDDP showed a more pronounced accumulation in mouse brain microvascular endothelial cells (bEnd.3) than BPD via an active transport process. The increased cellular uptake of PDDP was proven to be mediated by putative pyrilamine cationic transporters. Following intravenous administration, the concentration of PDDP in the brain was 269.28-fold and 6.41-fold higher than that of L-DOPA and BPD at 5 min, respectively. Additionally, PDDP effectively attenuated the striatum lesion caused by 6-hydroxydopamine (6-OHDA) in rats. More importantly, PDDP presented antioxidant and antiapoptotic effects on 6-OHDA-induced toxicity in human neuroblastoma cells (SH-SY5Y). Thus, N,N-dimethyl amino group-based PDDP represents an effective and safe treatment for PD.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Dopamin -hydrochlorid
Sigma-Aldrich
3,4-Dihydroxy-L-Phenylalanin, ≥98% (TLC)
Supelco
Levodopa, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Dopamin -hydrochlorid, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Levodopa, United States Pharmacopeia (USP) Reference Standard
Dopamin -hydrochlorid, European Pharmacopoeia (EP) Reference Standard
Levodopa, European Pharmacopoeia (EP) Reference Standard