Merck
  • Assessment of the optic disc morphology using spectral-domain optical coherence tomography and scanning laser ophthalmoscopy.
Assessment of the optic disc morphology using spectral-domain optical coherence tomography and scanning laser ophthalmoscopy.
BioMed research international (2014-08-12)
Pilar Calvo, Antonio Ferreras, Beatriz Abadia, Mirian Ara, Michele Figus, Luis E Pablo, Paolo Frezzotti
ZUSAMMENFASSUNG
To compare the equivalent optic nerve head (OHN) parameters obtained with confocal scanning laser ophthalmoscopy (HRT3) and spectral-domain optical coherence tomography (OCT) in healthy and glaucoma patients. One hundred and eighty-two consecutive healthy subjects and 156 patients with open-angle glaucoma were divided into 2 groups according to intraocular pressure and visual field outcomes. All participants underwent imaging of the ONH with the HRT3 and the Cirrus OCT. The ONH parameters and the receiver operating characteristic (ROC) curves were compared between both groups. Mean age did not differ between the normal and glaucoma groups (59.55 ± 9.7 years and 61.05 ± 9.4 years, resp.; P = 0.15). Rim area, average cup-to-disc (C/D) ratio, vertical C/D ratio, and cup volume were different between both instruments (P < 0.001). All equivalent ONH parameters, except disc area, were different between both groups (P < 0.001). The best areas under the ROC curve were observed for vertical C/D ratio (0.980 for OCT and 0.942 for HRT3; P = 0.11). Sensitivities at 95% fixed-specificities of OCT parameters were higher than those of HRT3. Equivalent ONH parameters of Cirrus OCT and HRT3 are different and cannot be used interchangeably. ONH parameters measured with OCT yielded a slightly better diagnostic performance.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Tropicamid, solid
Tropicamid, European Pharmacopoeia (EP) Reference Standard