Merck
  • p62/SQSTM1 is involved in cisplatin resistance in human ovarian cancer cells via the Keap1-Nrf2-ARE system.
p62/SQSTM1 is involved in cisplatin resistance in human ovarian cancer cells via the Keap1-Nrf2-ARE system.
International journal of oncology (2014-10-02)
Meihui Xia, Huimei Yu, Shuang Gu, Ye Xu, Jing Su, Hongyan Li, Jinsong Kang, Manhua Cui
ZUSAMMENFASSUNG
The mechanisms underlying cisplatin resistance in tumors are not fully understood. Previous studies have reported that cellular resistance to oxidative stress is accompanied by resistance to cisplatin. However, the relationship between the resistance to oxidative stress and cisplatin drug resistance in human ovarian cancer cells (HOCCs) is not clear. Here, we reveal a critical role for the multifunctional protein p62/SQSTM1 in cisplatin resistance in human ovarian cancer cells (HOCCs). p62/SQSTM1 (sequestosome 1) plays important roles in cell differentiation, proliferation and as an antiapoptotic molecule. We found that cisplatin-resistant SKOV3/DDP cells express much higher levels of p62 than do cisplatin-sensitive SKOV3 cells. The protein p62 can activate the Keap1-Nrf2-ARE signaling pathway and induce the expression of antioxidant genes in SKOV3/DDP cells. Knockdown of p62 resensitizes SKOV3/DDP cells to cisplatin. Collectively, our data indicate that cisplatin resistance in HOCCs is partially attributable to their high expression of p62, which plays an important role in preventing ROS stress-induced apoptosis by regulating the Keap1-Nrf2-ARE signaling pathway.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Propidiumjodid, ≥94.0% (HPLC)
Sigma-Aldrich
o-Phenylendiamin, flaked, 99.5%
Sigma-Aldrich
cis-Diamminplatin(II)-dichlorid, crystalline
Sigma-Aldrich
Propidiumjodid -Lösung
Sigma-Aldrich
o-Phenylendiamin, Peroxidase substrate, ≥98.0%, powder
Sigma-Aldrich
Propidiumjodid, ≥94% (HPLC)
Cisplatin, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
cis-Diaminplatin(II)-dichlorid, ≥99.9% trace metals basis
Sigma-Aldrich
o-Phenylendiamin, tablet, 20 mg substrate per tablet
Sigma-Aldrich
o-Phenylendiamin, sublimed, ≥99%
Sigma-Aldrich
trans-Platin(II)diammindichlorid
Cisplatin Unreinheit A, European Pharmacopoeia (EP) Reference Standard
USP
Transplatin, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Anti-GTF2H1 antibody produced in rabbit, IgG fraction of antiserum