Merck
Multiwalled nanotubes formed by catanionic mixtures of drug amphiphiles.
ACS nano (2014-11-22)
Yi-An Lin, Andrew G Cheetham, Pengcheng Zhang, Yu-Chuan Ou, Yuguo Li, Guanshu Liu, Daniel Hermida-Merino, Ian W Hamley, Honggang Cui
ZUSAMMENFASSUNG
Mixing of oppositely charged amphiphilic molecules (catanionic mixing) offers an attractive strategy to produce morphologies different from those formed by individual molecules. We report here on the use of catanionic mixing of anticancer drug amphiphiles to construct multiwalled nanotubes containing a fixed and high drug loading. We found that the molecular mixing ratio, the solvent composition, the overall drug concentrations, as well as the molecular design of the studied amphiphiles are all important experimental parameters contributing to the tubular morphology. We believe these results demonstrate the remarkable potential that anticancer drugs could offer to self-assemble into discrete nanostructures and also provide important insight into the formation mechanism of nanotubes by catanionic mixtures. Our preliminary animal studies reveal that the CPT nanotubes show significantly prolonged retention time in the tumor site after intratumoral injection.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
1,1,1,3,3,3-Hexafluor-2-propanol, ≥99%
Supelco
1,1,1,3,3,3-Hexafluor-2-propanol, derivatization grade (GC derivatization), LiChropur, ≥99.8%