Merck
  • pDCs efficiently process synthetic long peptides to induce functional virus- and tumour-specific T-cell responses.
pDCs efficiently process synthetic long peptides to induce functional virus- and tumour-specific T-cell responses.
European journal of immunology (2014-07-22)
Caroline Aspord, Claire Leloup, Sabine Reche, Joel Plumas
ZUSAMMENFASSUNG
Robust cell-mediated immunity is required for immune control of tumours and protection from viral infections, with both CD4(+) and CD8(+) T cells playing a pivotal role. Synthetic long peptides (SLPs) represent an attractive way to induce such combined responses, as they contain both class I and class II epitopes. The ability of plasmacytoid dendritic cells (pDCs) to cross-present SLPs has not yet been investigated; yet, pDCs play a critical role in shaping immune responses and have emerged as novel vectors for immunotherapy. Using overlapping 15-mer peptide pools covering the entire sequence of CMVpp65 and MelA, representing a viral disease (cytomegalovirus, CMV) and a tumour (melanoma), respectively, we showed that human pDCs can effectively process SLPs. Our results demonstrated that pDCs potently cross-present virus- and tumour-derived SLPs and cross-prime broad-ranging, effective and long-lived CD4(+) and CD8(+) T-cell responses, triggering more efficient immune responses than short peptide loaded pDCs. This ability required intracellular processing by the proteasome and was enhanced by co-exposure to TLR7/9-L. Combining SLPs with pDCs represents a powerful immunotherapeutic strategy to elicit potent immune responses, which are required for clinical success in cancers and viral infections.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Glutaraldehyd -Lösung, Grade I, 25% in H2O, specially purified for use as an electron microscopy fixative
Sigma-Aldrich
Natriumpyruvat, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Natriumpyruvat, ReagentPlus®, ≥99%
Sigma-Aldrich
Glutaraldehyd -Lösung, Grade II, 25% in H2O
Sigma-Aldrich
Glutaraldehyd -Lösung, 50 wt. % in H2O
Sigma-Aldrich
Natriumpyruvat, powder, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Glutaraldehyd -Lösung, Grade I, 50% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Natriumpyruvat, Hybri-Max, powder, suitable for hybridoma
Sigma-Aldrich
Glutaraldehyd -Lösung, Grade I, 70% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Glutaraldehyd -Lösung, technical, ~50% in H2O (5.6 M)
Sigma-Aldrich
Glutarsäure-Dialdehyd -Lösung, 50 wt. % in H2O, FCC
Sigma-Aldrich
Glutaraldehyd -Lösung, Grade I, 8% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Glutaraldehyd -Lösung, 50% in H2O, suitable for photographic applications
Sigma-Aldrich
Natriumpyruvat, BioXtra, ≥99%
Sigma-Aldrich
Natriumpyruvat, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%