Merck
Serine catabolism regulates mitochondrial redox control during hypoxia.
Cancer discovery (2014-09-05)
Jiangbin Ye, Jing Fan, Sriram Venneti, Ying-Wooi Wan, Bruce R Pawel, Ji Zhang, Lydia W S Finley, Chao Lu, Tullia Lindsten, Justin R Cross, Guoliang Qing, Zhandong Liu, M Celeste Simon, Joshua D Rabinowitz, Craig B Thompson
ZUSAMMENFASSUNG
The de novo synthesis of the nonessential amino acid serine is often upregulated in cancer. In this study, we demonstrate that the serine catabolic enzyme, mitochondrial serine hydroxymethyltransferase (SHMT2), is induced when MYC-transformed cells are subjected to hypoxia. In mitochondria, SHMT2 can initiate the degradation of serine to CO2 and NH4+, resulting in net production of NADPH from NADP+. Knockdown of SHMT2 in MYC-dependent cells reduced cellular NADPH:NADP+ ratio, increased cellular reactive oxygen species, and triggered hypoxia-induced cell death. In vivo, SHMT2 suppression led to impaired tumor growth. In MYC-amplified neuroblastoma patient samples, there was a significant correlation between SHMT2 and hypoxia-inducible factor-1 α (HIF1α), and SHMT2 expression correlated with unfavorable patient prognosis. Together, these data demonstrate that mitochondrial serine catabolism supports tumor growth by maintaining mitochondrial redox balance and cell survival. In this study, we demonstrate that the mitochondrial enzyme SHMT2 is induced upon hypoxic stress and is critical for maintaining NADPH production and redox balance to support tumor cell survival and growth.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Essigsäure, glacial, ACS reagent, ≥99.7%
Sigma-Aldrich
Essigsäure, glacial, ReagentPlus®, ≥99%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Essigsäure, glacial, ≥99.99% trace metals basis
Sigma-Aldrich
Essigsäure -Lösung, suitable for HPLC
Sigma-Aldrich
L-Glutathion reduziert, suitable for cell culture, BioReagent, ≥98.0%, powder
Sigma-Aldrich
L-Glutathion reduziert, ≥98.0%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Absolute - Acetone free
USP
Methylalkohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Supelco
Methanol, analytical standard
Sigma-Aldrich
Tributylamin, ≥98.5%
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Essigsäure, suitable for luminescence, BioUltra, ≥99.5% (GC)
USP
Eisessig, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
5α-Androstan-17β-ol-3-on, ≥97.5%
Supelco
Glutathion, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Tributylamin, puriss. plus, ≥99.5% (GC)
Supelco
Essigsäure, analytical standard
Sigma-Aldrich
L-Glutathion reduziert, BioXtra, ≥98.0%
Sigma-Aldrich
Essigsäure, ≥99.5%, FCC, FG
Sigma-Aldrich
Essigsäure, natural, ≥99.5%, FG