Merck
Organ-specific distribution of chlorophyll-related compounds from dietary spinach in rabbits.
Indian journal of biochemistry & biophysics (2015-01-30)
Ching-Yun Hsu, Tsan-Huei Yeh, Meng-Yuan Huang, Shene-Pin Hu, Pi-Yu Chao, Chi-Ming Yang
ZUSAMMENFASSUNG
The distribution of chlorophyll-related compounds (CRCs) derived from dietary spinach was investigated in different organs the rabbits. The rabbits in the experimental group consumed 100 g of freeze-dried spinach powder after a 24 h fasting period and sacrificed 2, 4, 8, 12 and 24 h later and in the control group sacrificed after the 24 h fasting period. The main CRCs in the liver were found to be chlorophyll (Chl a) and b, chlorophyllide (Chlide) a and b, pheophytin (Phe) a and b and pheophorbide (Pho) a and b, which reached their peak values at 8 h post-feeding. The gallbladder contained mainly Chlide a and a', Pho a and a', Pho b and b', which peaked their values at 2 h post-feeding. Pho a and b were consistently observed in the blood and peaked at 12 h post-feeding. The earlier appearance of Chlide a', Pho a' and Pho b' in the gallbladder compared to the liver indicated that these CRCs were compartmentalized differently and might undergo the same type of vectorialized transport as characterized for the bile salts. Pho levels peaked later in the blood compared to the liver, suggesting that Pho might be released into the peripheral blood circulation from the liver. In conclusion, Chlide and Pho were the principal Chl metabolites in the rabbits. Our data may expand our understanding of the metabolism and biodistribution of CRCs in the human body. A number of biological functions, including anti-oxidation, anti-tumor and anti-aging have recently been attributed to CRCs, it will be interesting to explore, if the binding of Chlide and Pho to other nutrients or trace metal ions in the body mediate their biological functions.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Aceton, ACS reagent, ≥99.5%
Sigma-Aldrich
Aceton, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Diethylether, anhydrous, ACS reagent, ≥99.0%, contains BHT as inhibitor
Sigma-Aldrich
Aceton, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%
Sigma-Aldrich
Diethylether, suitable for HPLC, ≥99.9%, inhibitor-free
Sigma-Aldrich
Diethylether, ACS reagent, anhydrous, ≥99.0%, contains BHT as inhibitor
Sigma-Aldrich
Diethylether, contains 1 ppm BHT as inhibitor, anhydrous, ≥99.7%
Sigma-Aldrich
Aceton, ACS reagent, ≥99.5%
Supelco
Aceton, analytical standard
Sigma-Aldrich
Diethylether
Sigma-Aldrich
Aceton, histological grade, ≥99.5%
Sigma-Aldrich
Diethylether, reagent grade, ≥98%, contains ≤2% ethanol and ≤10ppm BHT as inhibitor
Sigma-Aldrich
Diethylether, ACS reagent, ≥98.0%, contains ≤2% ethanol and ≤10ppm BHT as inhibitor
Supelco
Aceton, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Aceton, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Stickstoff, ≥99.998%
Supelco
Diethylether, analytical standard
Sigma-Aldrich
Aceton, ≥99%, FCC, FG
Sigma-Aldrich
Aceton, natural, ≥97%
Sigma-Aldrich
Aceton, suitable for HPLC, ≥99.9%