Merck
  • When assay format matters: a case study on the evaluation of three assay formats to support a clinical pharmacokinetic study.
When assay format matters: a case study on the evaluation of three assay formats to support a clinical pharmacokinetic study.
The AAPS journal (2014-04-23)
Kun Peng, Dana Baker, Suzanne Brignoli, Janis Cabuhat, Saloumeh K Fischer
ZUSAMMENFASSUNG
Data generated using various immunoassay methods are an integral part of the development of protein therapeutics. These assays are used in clinical and preclinical studies to establish the pharmacokinetic (PK) and pharmacodynamic (PD) characteristics as well as to assess the immunogenicity properties of a therapeutic. PK assays measure therapeutic levels post-administration which is essential for understanding the effective dose and dose regimen for a therapeutic. Anti-OX40L is a fully humanized monoclonal antibody designed for the potential treatment of an autoimmune disease. The anti-OX40L human PK assay is required to be sensitive, robust, and precise. To address challenges due to assay sensitivity and reproducibility, as well as assay technology limitations, during development of the anti-OX40L human PK assay, three different assays, including an MSD-based electrochemiluminescence assay (ECLA), a fluorometric enzyme-linked immunosorbent assay (ELISA), and a colorimetric ELISA, were evaluated. The MSD-based assay was the most sensitive but posed risk of inter-well signal crosstalk. The fluorescence ELISA fell short on reproducibility. The colorimetric ELISA was ultimately chosen for supporting sample analysis. This paper presents characterization data obtained from each of these assay formats, challenges that were encountered in the development of the assay, and the rationale for selecting the ultimate assay format.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Phosphorsäure, ACS reagent, ≥85 wt. % in H2O
Sigma-Aldrich
Phosphorsäure, ACS reagent, ≥85 wt. % in H2O
Sigma-Aldrich
Phosphorsäure, 85 wt. % in H2O, 99.99% trace metals basis
Sigma-Aldrich
Phosphorsäure, crystalline, ≥99.999% trace metals basis
Sigma-Aldrich
Phosphorsäure -Lösung, 85 wt. % in H2O, FCC, FG
Sigma-Aldrich
Phosphorsäure, BioUltra, ≥85% (T)
Sigma-Aldrich
Phosphorsäure, ≥85 wt. % in H2O, ≥99.999% trace metals basis
Sigma-Aldrich
Phosphorsäure, BioReagent, suitable for insect cell culture, 85%
Sigma-Aldrich
3-[(3-Cholamidopropyl)dimethylammonio]-1-propansulfonat Hydrat, 98%
Sigma-Aldrich
Phosphorsäure -Lösung, suitable for NMR (reference standard), 85% in D2O (99.9 atom % D), NMR tube size 4.2 mm × 8 in. , WGS-5BL Coaxial NMR tube
Sigma-Aldrich
Phosphorsäure -Lösung, suitable for NMR (reference standard), 85% in D2O (99.9 atom % D), NMR tube size 5 mm × 8 in.
Sigma-Aldrich
Phosphorsäure -Lösung, suitable for NMR (reference standard), 85% in D2O (99.9 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Phosphorsäure-16O4 -Lösung, 70 wt. % in D2O, 99.9 atom % 16O