Merck
  • Capture of the circulating Plasmodium falciparum biomarker HRP2 in a multiplexed format, via a wearable skin patch.
Capture of the circulating Plasmodium falciparum biomarker HRP2 in a multiplexed format, via a wearable skin patch.
Analytical chemistry (2014-09-19)
Khai Tuck Lee, David A Muller, Jacob W Coffey, Kye J Robinson, James S McCarthy, Mark A F Kendall, Simon R Corrie
ZUSAMMENFASSUNG
Herein we demonstrate the use of a wearable device that can selectively capture two distinct circulating protein biomarkers (recombinant P. falciparum rPfHRP2 and total IgG) from the intradermal fluid of live mice in situ, for subsequent detection in vitro. The device comprises a microprojection array that, when applied to the skin, penetrates the outer skin layers to interface directly with intradermal fluid. Because of the complexity of the biological fluid being sampled, we investigated the effects of solution conditions on the attachment of capture antibodies, to optimize the assay detection limit both in vitro and on live mice. For detection of the target antigen diluted in 20% serum, immobilization conditions favoring high antibody surface density (low pH, low ionic strength) resulted in 100-fold greater sensitivity in comparison to standard conditions, yielding a detection limit equivalent to the plate enzyme-linked immunosorbent assay (ELISA). We also show that blocking the device surface to reduce nonspecific adsorption of target analyte and host proteins does not significantly change sensitivity. After injecting mice with rPfHRP2 via the tail vein, we compared analyte levels in both plasma and skin biopsies (cross-sectional area same as the microprojection array), observing that skin samples contained the equivalent of ∼8 μL of analyte-containing plasma. We then applied the arrays to mice, showing that surfaces coated with a high density of antibodies captured a significant amount of the rPfHRP2 target while the standard surface showed no capture in comparison to the negative control. Next, we applied a triplex device to both control and rPfHRP2-treated mice, simultaneously capturing rPfHRP2 and total IgG (as a positive control for skin penetration) in comparison to a negative control device. We conclude that such devices can be used to capture clinically relevant, circulating protein biomarkers of infectious disease via the skin, with potential applications as a minimally invasive and lab-free biomarker detection platform.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Phosphorsäure, ACS reagent, ≥85 wt. % in H2O
Sigma-Aldrich
Phosphorsäure, ACS reagent, ≥85 wt. % in H2O
Sigma-Aldrich
Phosphorsäure, 85 wt. % in H2O, 99.99% trace metals basis
Sigma-Aldrich
Phosphorsäure, crystalline, ≥99.999% trace metals basis
Sigma-Aldrich
Phosphorsäure -Lösung, 85 wt. % in H2O, FCC, FG
Sigma-Aldrich
Phosphorsäure, BioUltra, ≥85% (T)
Sigma-Aldrich
Phosphorsäure, ≥85 wt. % in H2O, ≥99.999% trace metals basis
Sigma-Aldrich
Phosphorsäure, BioReagent, suitable for insect cell culture, 85%
Sigma-Aldrich
Phosphorsäure -Lösung, suitable for NMR (reference standard), 85% in D2O (99.9 atom % D), NMR tube size 4.2 mm × 8 in. , WGS-5BL Coaxial NMR tube
Sigma-Aldrich
Phosphorsäure -Lösung, suitable for NMR (reference standard), 85% in D2O (99.9 atom % D), NMR tube size 5 mm × 8 in.
Sigma-Aldrich
Phosphorsäure -Lösung, suitable for NMR (reference standard), 85% in D2O (99.9 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Phosphorsäure-16O4 -Lösung, 70 wt. % in D2O, 99.9 atom % 16O