Merck
Osteopontin facilitates ultraviolet B-induced squamous cell carcinoma development.
Journal of dermatological science (2014-06-04)
Pi-Ling Chang, Yu-Hua Hsieh, Chao-Cheng Wang, M Margaret Juliana, Yuko Tsuruta, Laura Timares, Craig Elmets, Kang-Jey Ho
ZUSAMMENFASSUNG
Osteopontin (OPN) is a matricellular glycoprotein that is markedly expressed in cutaneous squamous cell carcinomas (cSCCs) and in actinic keratoses implicating its role in photocarcinogenesis. To determine whether OPN facilitates the development of cSCC and its function. cSCCs development was compared between wild-type (WT) and OPN-null mice subjected to UVB irradiation for 43 weeks. UVB-induced OPN expression was determined by Western blot, immunoprecipitation, ELISA, and semi-quantitative RT-PCR. Epidermal layer and TUNEL analyses assessed if OPN mediates UVB-induced epidermal hyperplasia or suppresses UVB-induced apoptosis of basal keratinocytes, respectively. In vitro experiments determined whether OPN enhances cell survival of UVB-induced apoptosis and its potential mechanisms. Immunohistochemical analyses of epidermis assessed the expression of CD44 and focal adhesion kinase (FAK), molecules that mediate OPN survival function. Compared to female WT mice, OPN-null mice did not develop cSCCs. UVB irradiation stimulated OPN protein expression in the dorsal skin by 11h and remains high at 24-48h. OPN did not mediate UVB-induced epidermal hyperplasia; instead, it protected basal keratinocytes from undergoing apoptosis upon UVB exposure. Likewise, the addition of OPN suppressed UVB-induced OPN-null cSCC cell apoptosis, the activation of caspase-9 activity, and increased phosphorylation of FAK at Y397. Furthermore, the expression of CD44 and FAK in WT mice epidermis was greater than that of OPN-null mice prior to and during early acute UVB exposure. These data support the hypothesis that chronic UVB-induced OPN expression protects the survival of initiated basal keratinocytes and, consequently, facilitates cSCC develop.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Calciumchlorid -Lösung, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Calciumchlorid, anhydrous, BioReagent, suitable for insect cell culture, suitable for plant cell culture, ≥96.0%
Sigma-Aldrich
Calciumchlorid, powder, 99.99% trace metals basis
Sigma-Aldrich
Calciumchlorid Dihydrat
Sigma-Aldrich
Calciumchlorid, AnhydroBeads, −10 mesh, ≥99.9% trace metals basis
Supelco
Calcium Ionenlösung für ISE, 0.1 M Ca, analytical standard (for ion-selective electrodes)
Sigma-Aldrich
Calciumchlorid, AnhydroBeads, −10 mesh, ≥99.99% trace metals basis