Merck
The role of non-placental signals in the adaptation of islets to pregnancy.
Hormone and metabolic research = Hormon- und Stoffwechselforschung = Hormones et metabolisme (2014-12-17)
R Drynda, C J Peters, P M Jones, J E Bowe
ZUSAMMENFASSUNG
It is well established that the maternal β-cell mass increases during pregnancy in both humans and rodents to compensate insulin resistance and increased metabolic demand, and rapidly returns to normal levels post-partum. However, the mechanisms underlying this adaptation are not well understood. It is established that this process is driven partly by placental signals, but the contribution of non-placental signals is still unclear. This study aimed to differentiate between the role of placental and non-placental signals in regulating the β-cell mass and glucose homeostasis during and after pregnancy. Pseudopregnant, pregnant and lactating mice were used to study the effects of maternal hormones on β-cell function during early pregnancy, mid-to-late pregnancy and post-partum, respectively. Pseudopregnant mice, with circulating hormone levels mirroring those during pregnancy but lacking placental signals, had significantly increased β-cell proliferation compared to non-pregnant controls but no change in glucose homeostasis, suggesting a role for non-placental hormones in increasing β-cell mass. The rate of β-cell proliferation rate dropped immediately after parturition, but lactating mice still had a significantly higher rate of β-cell proliferation compared to non-lactating post-partum mice, suggesting that lactation-related hormones play a role in the controlled involution of β-cell mass post-partum. These results implicate a role for both non-placental and placental signals in regulating β-cell mass during and after pregnancy.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Ethyl alcohol, Pure 200 proof, Molecular Biology
Sigma-Aldrich
Salzsäure, ACS reagent, 37%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Salzsäure, ACS reagent, 37%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Sigma-Aldrich
Chlorwasserstoff -Lösung, 4.0 M in dioxane
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure 190 proof, for molecular biology
Sigma-Aldrich
Salzsäure -Lösung, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Salzsäure, meets analytical specification of Ph. Eur., BP, NF, fuming, 36.5-38%
Sigma-Aldrich
Salzsäure, 37 wt. % in H2O, 99.999% trace metals basis
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Chlorwasserstoff -Lösung, 2.0 M in diethyl ether
Sigma-Aldrich
Thymidin, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethanol, BioUltra, Molecular Biology, ≥99.8%, (absolute alcohol, without additive, A15 o1)
Sigma-Aldrich
Salzsäure, 36.5-38.0%, BioReagent, Molecular Biology
Supelco
Salzsäure -Lösung, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Thymidin, ≥99%
Supelco
Ethanol, wasserfrei, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Chlorwasserstoff -Lösung, 1.0 M in diethyl ether
Supelco
Ethanol, standard for GC
Sigma-Aldrich
Ethanol, purum, fine spirit, denaturated with 4.8% methanol, F25 METHYL1, ~96% (based on denaturant-free substance)
Sigma-Aldrich
Salzsäure -Lösung, for amino acid analysis, ~6 M in H2O
Sigma-Aldrich
Chlorwasserstoff, ReagentPlus®, ≥99%
Sigma-Aldrich
Chlorwasserstoff -Lösung, 3 M in cyclopentyl methyl ether (CPME)
Supelco
Chlorwasserstoff – Methanol -Lösung, ~1.25 m HCl (T), derivatization grade (GC derivatization), LiChropur
Sigma-Aldrich
Salzsäure -Lösung, 32 wt. % in H2O, FCC
Sigma-Aldrich
5-Brom-2′-deoxyuridin, BioUltra, ≥99%
Sigma-Aldrich
Ethanol
Supelco
Ethanol -Lösung, certified reference material, 2000 μg/mL in methanol