Merck
Nicotine reduces distraction under low perceptual load.
Psychopharmacology (2014-10-12)
Oliver Behler, Thomas P K Breckel, Christiane M Thiel
ZUSAMMENFASSUNG
Several studies provide evidence that nicotine alleviates the detrimental effects of distracting sensory stimuli. It is been suggested that nicotine may either act as a stimulus filter that prevents irrelevant stimuli entering awareness or by enhancing the attentional focus to relevant stimuli via a boost in processing capacity. To differentiate between these two accounts, we administered nicotine to healthy non-smokers and investigated distractor interference in a visual search task with low and high perceptual load to tax processing capacity. Thirty healthy non-smokers received either 7 mg transdermal nicotine or a matched placebo in a double blind within subject design 1 h prior to performing the visual search task with different fixation distractors. Nicotine reduced interference of incongruent distractors, but only under low-load conditions, where distractor effects were large. No effects of nicotine were observed under high-load conditions. Highly distractible subjects showed the largest effects of nicotine. The findings suggest that nicotine acts primarily as a stimulus filter that prevents irrelevant stimuli from entering awareness in situations of high distractor interference.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
(−)-Nikotin, ≥99% (GC), liquid
Sigma-Aldrich
(±)-Nikotin, ≥99% (TLC), liquid
Supelco
(−)-Nikotin, PESTANAL®, analytical standard