Merck
  • A novel pH-responsive interpolyelectrolyte hydrogel complex for the oral delivery of levodopa. Part II: characterization and formulation of an IPEC-based tablet matrix.
A novel pH-responsive interpolyelectrolyte hydrogel complex for the oral delivery of levodopa. Part II: characterization and formulation of an IPEC-based tablet matrix.
Journal of biomedical materials research. Part A (2014-06-10)
Ndidi C Ngwuluka, Yahya E Choonara, Pradeep Kumar, Lisa C du Toit, Riaz A Khan, Viness Pillay
ZUSAMMENFASSUNG
This study was undertaken in order to apply a synthesized interpolyelectrolyte complex (IPEC) of polymethacrylate and carboxymethylcellulose as a controlled release oral tablet matrix for the delivery of the model neuroactive drug levodopa. The IPEC (synthesized in Part I of this work) was characterized by techniques such as Fourier Transform Infra-Red (FTIR) spectroscopy, Differential Scanning Calorimetry (DSC), Advanced DSC (ADSC), and Scanning Electron Microscopy (SEM). The tablet matrices were formulated and characterized for their drug delivery properties and in vitro drug release. FTIR confirmed the interaction between the two polymers. The IPEC composite generated tablet matrices with a hardness ranging from 19.152-27.590 N/mm and a matrix resilience ranging between 42 and 46%. An IPEC of polymethacrylate and carboxymethylcellulose was indeed an improvement on the inherent properties of the native polymers providing a biomaterial with the ability to release poorly soluble drugs such as levodopa at a constant rate over a prolonged period of time.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Salzsäure, ACS reagent, 37%
Sigma-Aldrich
Kaliumphosphat, ACS reagent, ≥99.0%
Sigma-Aldrich
Phosphorsäure, ACS reagent, ≥85 wt. % in H2O
Sigma-Aldrich
Salzsäure, ACS reagent, 37%
Sigma-Aldrich
Chlorwasserstoff -Lösung, 4.0 M in dioxane
Sigma-Aldrich
Phosphorsäure, ACS reagent, ≥85 wt. % in H2O
Sigma-Aldrich
Phosphorsäure, 85 wt. % in H2O, 99.99% trace metals basis
Sigma-Aldrich
Kaliumphosphat, powder, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0%
Sigma-Aldrich
Salzsäure -Lösung, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Salzsäure, meets analytical specification of Ph. Eur., BP, NF, fuming, 36.5-38%
Sigma-Aldrich
Salzsäure, 37 wt. % in H2O, 99.999% trace metals basis
Sigma-Aldrich
Chlorwasserstoff -Lösung, 2.0 M in diethyl ether
Sigma-Aldrich
Salzsäure, 36.5-38.0%, BioReagent, Molecular Biology
Sigma-Aldrich
3,4-Dihydroxy-L-Phenylalanin, ≥98% (TLC)
Sigma-Aldrich
Phosphorsäure, crystalline, ≥99.999% trace metals basis
Supelco
Salzsäure -Lösung, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Kaliumphosphat, ReagentPlus®
Sigma-Aldrich
Kaliumphosphat, Molecular Biology, ≥98.0%
Sigma-Aldrich
Phosphorsäure -Lösung, 85 wt. % in H2O, FCC, FG
Sigma-Aldrich
Chlorwasserstoff -Lösung, 1.0 M in diethyl ether
Sigma-Aldrich
Salzsäure -Lösung, for amino acid analysis, ~6 M in H2O
Sigma-Aldrich
Phosphorsäure, BioUltra, ≥85% (T)
Sigma-Aldrich
Kalium, chunks (in mineral oil), 98% trace metals basis
Sigma-Aldrich
Chlorwasserstoff, ReagentPlus®, ≥99%
Sigma-Aldrich
Chlorwasserstoff -Lösung, 3 M in cyclopentyl methyl ether (CPME)
Supelco
Chlorwasserstoff – Methanol -Lösung, ~1.25 m HCl (T), derivatization grade (GC derivatization), LiChropur
Supelco
Kaliumphosphat, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Kaliumphosphat, BioUltra, Molecular Biology, anhydrous, ≥99.5% (T)
Sigma-Aldrich
Kaliumhydrid, 30 wt % dispersion in mineral oil
Sigma-Aldrich
Salzsäure -Lösung, 32 wt. % in H2O, FCC