Merck
Chemokine-mediated robust augmentation of liver engraftment: a novel approach.
Stem cells translational medicine (2014-12-05)
Meghnad Joshi, Mihai Oltean, Pradeep B Patil, David Hallberg, Marika Kleman, Jan Holgersson, Michael Olausson, Suchitra Sumitran-Holgersson
ZUSAMMENFASSUNG
Effective repopulation of the liver is essential for successful clinical hepatocyte transplantation. The objective was to improve repopulation of the liver with human hepatocytes using chemokines. We used flow cytometry and immunohistochemistry assays to identify commonly expressed chemokine receptors on human fetal and adult hepatocytes. The migratory capacity of the cells to various chemokines was tested. For in vivo studies, we used a nude mouse model of partial hepatectomy followed by intraparenchymal injections of chemokine ligands at various concentrations. Human fetal liver cells transformed with human telomerase reverse transcriptase were used for intrasplenic cell transplantation. Repopulation and functionality were assessed 4 weeks after transplantation. The receptor CXCR3 was commonly expressed on both fetal and adult hepatocytes. Both cell types migrated efficiently toward corresponding CXC chemokine ligands 9, 10, and 11. In vivo, animals injected with recombinant chemokines showed the highest cell engraftment compared with controls (p<.05). The engrafted cells expressed several human hepatic markers such as cytokeratin 8 and 18 and albumin as well as transferrin, UGT1A1, hepatocyte nuclear factor (1α, 1β, and 4α), cytochrome CYP3A1, CCAAT/enhancer binding protein (α and β), and human albumin compared with controls. No inflammatory cells were detected in the livers at 4 weeks after transplantation. The improved repopulation of transplanted cells is likely a function of the chemokines to mediate cell homing and retention in the injured liver and might be an attractive strategy to augment repopulation of transplanted hepatocytes in vivo.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Ammoniak -Lösung, 7 N in methanol
Sigma-Aldrich
L-Glutamin, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
Natriumpyruvat, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
L-Glutamin, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Natriumpyruvat, ReagentPlus®, ≥99%
SAFC
L-Glutamin
Sigma-Aldrich
Ammoniak -Lösung, 2.0 M in ethanol
Sigma-Aldrich
Ammoniak, anhydrous, ≥99.98%
Sigma-Aldrich
Ammoniak -Lösung, 0.4 M in dioxane
Sigma-Aldrich
Ammoniak-Assay-Kit, sufficient for 100 assays
Sigma-Aldrich
Ammoniak -Lösung, 4 M in methanol
Sigma-Aldrich
Ammoniak -Lösung, 0.4 M in THF
Sigma-Aldrich
Phenolrot, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Phenolrot, ACS reagent
Sigma-Aldrich
Natriumpyruvat, powder, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
L-Glutamin, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
Ammoniak -Lösung, 2.0 M in isopropanol
Sigma-Aldrich
Natriumpyruvat, Hybri-Max, powder, suitable for hybridoma
Sigma-Aldrich
QCM Chemotaxis-Zellmigrationsassay, 24-Well (8 µm), kolorimetrisch, The QCM 24-well Migration Assay is ideal for the study of chemotaxis cell migration. The assay uses a 24-well plate with an 8 micron pore size, with colorimetric detection.
Sigma-Aldrich
L-Glutamin, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
L-Glutamin
Sigma-Aldrich
Ammoniak, puriss., anhydrous, ≥99.95%
Sigma-Aldrich
Natriumpyruvat, BioXtra, ≥99%
Supelco
L-Glutamin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Natriumpyruvat, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%
Supelco
L-Glutamin, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Ammoniak-14N, 99.99 atom % 14N