Merck
  • AMSH-mediated deubiquitination of Cx43 regulates internalization and degradation of gap junctions.
AMSH-mediated deubiquitination of Cx43 regulates internalization and degradation of gap junctions.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology (2014-07-30)
Teresa M Ribeiro-Rodrigues, Steve Catarino, Carla Marques, João V Ferreira, Tânia Martins-Marques, Paulo Pereira, Henrique Girão
ZUSAMMENFASSUNG
Gap junctions (GJs) are specialized cell-cell contacts formed by connexins (Cxs), which provide direct intercellular communication between eukaryotic cells. Although Cx43 has long been known to be a substrate for ubiquitination, the reversal of this modification by deubiquitylases (DUBs) has never been described. Here we report that the DUB-associated molecule with the SH3 domain of STAM (AMSH) interacts with Cx43 and mediates its deubiquitination. In this study, we demonstrate that Cx43 is modified with lysine 63-linked polyubiquitin chains and that these increase the interaction between Cx43 and AMSH. We also show that AMSH is recruited to GJ plaque sites at the plasma membrane, where it mediates the deubiquitination of Cx43. Using siRNA depletion or overexpression of a catalytically inactive mutant of AMSH, we show that by decreasing Cx43 deubiquitination, both the internalization and degradation rate of Cx43 are increased. Overall, these data strongly suggest that AMSH-mediated deubiquitination of Cx43 protects GJs from degradation.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Phorbol-12-myristat-13-acetat, ≥99% (TLC), film or powder
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
PMA, for use in molecular biology applications, ≥99% (HPLC), Molecular Biology
Sigma-Aldrich
Actidion, from microbial, ≥94% (TLC)
Sigma-Aldrich
Cycloheximid -Lösung, Ready-Made Solution, microbial, 100 mg/mL in DMSO, Suitable for cell culture
Sigma-Aldrich
Cycloheximid, ≥95% (HPLC)
Sigma-Aldrich
Actidion, Biotechnology Performance Certified
Sigma-Aldrich
Phorbol-12-myristat-13-acetat, synthetic, ≥98.0% (TLC)
Millipore
Cycloheximid -Lösung, 0.1%, suitable for microbiology
Supelco
Actidion, PESTANAL®, analytical standard
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Stambp
Sigma-Aldrich
MISSION® esiRNA, targeting human STAMBP