Merck
  • Hepatoprotective effect of silymarin on di(2-ethylhexyl)phthalate (DEHP) induced injury in liver FL83B cells.
Hepatoprotective effect of silymarin on di(2-ethylhexyl)phthalate (DEHP) induced injury in liver FL83B cells.
Environmental toxicology and pharmacology (2014-06-18)
Diana Lo, Yuh-Tai Wang, Ming-Chang Wu
ZUSAMMENFASSUNG
Di(2-ethylhexyl)phthalate (DEHP), is a toxic environmental pollutant in our life which can contaminate air, water, and soil. The hepatoprotective effect of silymarin on DEHP-induced injury in FL83B mouse liver cells was investigated by analyzing the cell viability, lactate dehydrogenase (LDH), alanine aminotransferase (ALT), cell cycle arrest, and cell morphology. The results revealed that cell viability decreased while released LDH and ALT increased with the increase of DEHP concentrations. Moreover, cell population of sub-G1 and S phase increased as the concentrations of DEHP increased. Silymarin at 25 μM achieved the highest hepatoprotective effect and exhibited 79% cell viability while only 46% cell viability was found in DEHP injured control. It was also found to reduce LDH release and cell populations of sub-G1 and S phase. Therefore, silymarin could ameliorate DEHP-induced injury and have potential to be further developed as a natural ingredient of health food against phthalate plasticizers induced liver injury.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
L-Glutamin, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
Propidiumjodid, ≥94.0% (HPLC)
Sigma-Aldrich
L-Glutamin, ReagentPlus®, ≥99% (HPLC)
SAFC
L-Glutamin
Sigma-Aldrich
Propidiumjodid -Lösung
Sigma-Aldrich
L-Glutamin, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
Propidiumjodid, ≥94% (HPLC)
Supelco
Phthalsäure-bis-2-ethylhexylester, PESTANAL®, analytical standard
Sigma-Aldrich
L-Glutamin, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
L-Glutamin
Supelco
Phthalsäure-bis-2-ethylhexylester, Selectophore
Supelco
Phthalsäure-bis-2-ethylhexylester, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
L-Glutamin, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
L-Glutamin, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Kunststoffzusatzstoff 01, European Pharmacopoeia (EP) Reference Standard