Merck
  • PGRN induces impaired insulin sensitivity and defective autophagy in hepatic insulin resistance.
PGRN induces impaired insulin sensitivity and defective autophagy in hepatic insulin resistance.
Molecular endocrinology (Baltimore, Md.) (2015-02-11)
Jiali Liu, Huixia Li, Bo Zhou, Lin Xu, Xiaomin Kang, Wei Yang, Shufang Wu, Hongzhi Sun
ZUSAMMENFASSUNG
Progranulin (PGRN) has recently emerged as an important regulator for glucose metabolism and insulin sensitivity. However, the underlying mechanisms of PGRN in the regulation of insulin sensitivity and autophagy remain elusive. In this study, we aimed to address the direct effects of PGRN in vivo and to evaluate the potential interaction of impaired insulin sensitivity and autophagic disorders in hepatic insulin resistance. We found that mice treated with PGRN for 21 days exhibited the impaired glucose tolerance and insulin tolerance and hepatic autophagy imbalance as well as defective insulin signaling. Furthermore, treatment of mice with TNF receptor (TNFR)-1 blocking peptide-Fc, a TNFR1 blocking peptide-Fc fusion protein to competitively block the interaction of PGRN and TNFR1, resulted in the restoration of systemic insulin sensitivity and the recovery of autophagy and insulin signaling in liver. Consistent with these findings in vivo, we also observed that PGRN treatment induced defective autophagy and impaired insulin signaling in hepatocytes, with such effects being drastically nullified by the addition of TNFR1 blocking peptide -Fc or TNFR1-small interference RNA via the TNFR1-nuclear factor-κB-dependent manner, indicating the causative role of PGRN in hepatic insulin resistance. In conclusion, our findings supported the notion that PGRN is a key regulator of hepatic insulin resistance and that PGRN may mediate its effects, at least in part, by inducing defective autophagy via TNFR1/nuclear factor-κB.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Oleinsäure, technical grade, 90%
Sigma-Aldrich
Oleinsäure, ≥99% (GC)
Sigma-Aldrich
Oleinsäure, suitable for cell culture, BioReagent
Sigma-Aldrich
Palmitinsäure, BioXtra, ≥99%
Sigma-Aldrich
Oleinsäure, natural, FCC
Sigma-Aldrich
Oleinsäure, meets analytical specification of Ph, Eur., 65.0-88.0% (GC)
Supelco
Oleinsäure, analytical standard
Supelco
Oleinsäure, Selectophore, ≥99%
Oleinsäure, European Pharmacopoeia (EP) Reference Standard