Merck
Polymer fullerene solution phase behaviour and film formation pathways.
Soft matter (2015-03-06)
Rajeev Dattani, João T Cabral
ZUSAMMENFASSUNG
We report the phase behaviour of polymer/fullerene/solvent ternary mixtures and its consequence for the morphology of the resulting composite thin films. We focus particularly on solutions of polystyrene (PS), C60 fullerene and toluene, which are examined by static and dynamic light scattering, and films obtained from various solution ages and thermal annealing conditions, using atomic force and light microscopy. Unexpectedly, the solution phase behaviour below the polymer overlap concentration, c*, is found to be described by a simple excluded volume argument (occupied by the polymer chains) and the neat C60/solvent miscibility. Scaling consistent with full exclusion is found when the miscibility of the fullerene in the solvent is much lower than that of the polymer, giving way to partial exclusion with more soluble fullerenes (phenyl-C61-butyric acid methyl ester, PCBM) and a less asymmetric solvent (chlorobenzene), employed in photovoltaic devices. Spun cast and drop cast films were prepared from PS/C60/toluene solutions across the phase diagram to yield an identical PS/C60 composition and film thickness, resulting in qualitatively different morphologies in agreement with our measured solution phase boundaries. Our findings are relevant to the solution processing of polymer/fullerene composites (including organic photovoltaic devices), which generally require effective solubilisation of fullerene derivatives and polymer pairs in this concentration range, and the design of well-defined thin film morphologies.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Toluol, ACS reagent, ≥99.5%
Sigma-Aldrich
Toluol, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%
Sigma-Aldrich
Toluol, anhydrous, 99.8%
Sigma-Aldrich
Chlorbenzol, anhydrous, 99.8%
Sigma-Aldrich
Chlorbenzol, suitable for HPLC, 99.9%
Sigma-Aldrich
Chlorbenzol, ACS reagent, ≥99.5%
Sigma-Aldrich
Toluol, ACS reagent, ≥99.5%
Supelco
Toluol, analytical standard
Supelco
Toluol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Toluol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Toluol, ACS reagent, ≥99.5%
Supelco
Chlorbenzol, analytical standard