Merck
Nitrosylation of c heme in cd(1)-nitrite reductase is enhanced during catalysis.
Biochemical and biophysical research communications (2014-08-15)
Serena Rinaldo, Giorgio Giardina, Francesca Cutruzzolà
ZUSAMMENFASSUNG
The reduction of nitrite into nitric oxide (NO) in denitrifying bacteria is catalyzed by nitrite reductase. In several species, this enzyme is a heme-containing protein with one c heme and one d1 heme per monomer (cd1NiR), encoded by the nirS gene. For many years, the evidence of a link between NO and this hemeprotein represented a paradox, given that NO was known to tightly bind and, possibly, inhibit hemeproteins, including cd1NiRs. It is now established that, during catalysis, cd1NiRs diverge from "canonical" hemeproteins, since the product NO rapidly dissociates from the ferrous d1 heme, which, in turn, displays a peculiar "low" affinity for NO (KD=0.11 μM at pH 7.0). It has been also previously shown that the c heme reacts with NO at acidic pH but c heme nitrosylation was not extensively investigated, given that in cd1NiR it was considered a side reaction, rather than a genuine process controlling catalysis. The spectroscopic study of the reaction of cd1NiR and its semi-apo derivative (containing the sole c heme) with NO reported here shows that c heme nitrosylation is enhanced during catalysis; this evidence has been discussed in order to assess the potential of c heme nitrosylation as a regulatory process, as observed for cytochrome c nitrosylation in mammalian mitochondria.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Natriumnitrit, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Natriumnitrit, ACS reagent, ≥97.0%
Sigma-Aldrich
Natriumnitrit, 99.999% trace metals basis
Supelco
Natriumnitrit, analytical standard
Supelco
Nitrit Ionen-Standardlösung, for ion-selective electrodes, 0.1 M NO2-
Sigma-Aldrich
Natriumnitrit, anhydrous, Redi-Dri, ReagentPlus®, ≥99.0%