Merck
Evaluation of a biofilm formation by Desulfovibrio fairfieldensis on titanium implants.
Letters in applied microbiology (2014-11-29)
F P A Jorand, S Debuy, S F Kamagate, M Engels-Deutsch
ZUSAMMENFASSUNG
The aim of this study was to assess the capabilities of Desulfovibrio fairfieldensis to colonize the grade 4 titanium coupons (modSLA) used in dental implants. The effect of ampicillin, which is known to be a poorly penetrating agent in the matrix biofilm, was also compared with planktonic and sessile cells. The modSLA colonization by bacteria in KNO3 (0.05 mol l(-1)) and culture media (DSM 63 and fetal bovine serum) was determined by direct cell counts and field emission electronic microscopy. The surface of titanium (Ti) coupons was characterized by scanning electron microscopy and by Raman spectroscopy. Cells, mainly located in surface pores of modSLA coupons, appeared to be wrapped in a polymeric-like structure. The initial apparent rates of adhesion varied from 3 × 10(6) to 30 × 10(6) cells cm(-2) h(-1), and a plateau was reached at 1 day, regardless of the incubation medium. No cells have significantly adhered to polished Ti, and a minority was found on massive Ti. Finally, cells trapped on the modSLA surface were not lysed by ampicillin contrary to planktonic cells. Des. fairfieldensis is therefore able to colonize the rough surface of modSLA implant through a physical trapping in the microporosity of the surface, where they can produce a biofilm-like structure to improve their resistance to ampicillin. Desulfovibrio fairfieldensis is one of the most relevant sulphate-reducing bacteria of the human oral cavity suspected to be involved in peri-implantitis and implant corrosion. This study demonstrates for the first time that Des. fairfieldensis is able to initiate the formation of a biofilm-like structure on the microstructured titanium coupons used in dental implants and that it improves its resistance to antibiotic treatment. It gives new insight to understand the capacity of this opportunistic pathogen to colonize implant surfaces and to resist to biocide treatments.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Kaliumnitrat, ACS reagent, ≥99.0%
Sigma-Aldrich
Formaldehyd -Lösung, Molecular Biology, 36.5-38% in H2O
Sigma-Aldrich
Formaldehyd -Lösung, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
SAFC
Formaldehyd -Lösung, contains 10-15% methanol as stabilizer, 37 wt. % in H2O
Sigma-Aldrich
Kaliumnitrat, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99%
Sigma-Aldrich
Kaliumnitrat, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Formaldehyd -Lösung, Molecular Biology, BioReagent, ≥36.0% in H2O (T)
Sigma-Aldrich
Kaliumnitrat, BioReagent, suitable for cell culture, suitable for plant cell culture
Sigma-Aldrich
Formaldehyd -Lösung, meets analytical specification of USP, ≥34.5 wt. %
Supelco
Formaldehyd -Lösung, stabilized with methanol, ~37 wt. % in H2O, certified reference material
Sigma-Aldrich
Formaldehyd -Lösung, tested according to Ph. Eur.
Sigma-Aldrich
Kaliumnitrat, 99.999% trace metals basis
Sigma-Aldrich
Kaliumnitrat, BioXtra, ≥99.0%
Sigma-Aldrich
Kaliumnitrat, 99.99% trace metals basis
Supelco
Mettler-Toledo-Kalibriersubstanz ME 51143095, Kaliumnitrat, traceable to primary standards (LGC)
Sigma-Aldrich
Formaldehyd-12C -Lösung, 20% in H2O, 99.9 atom % 12C