Merck
  • Catalytic hydropyrolysis of microalgae: influence of operating variables on the formation and composition of bio-oil.
Catalytic hydropyrolysis of microalgae: influence of operating variables on the formation and composition of bio-oil.
Bioresource technology (2014-08-28)
Zhoufan Chang, Peigao Duan, Yuping Xu
ZUSAMMENFASSUNG
Catalytic hydropyrolysis of microalgae has been studied by using a batch reactor. Nine different heterogenous catalysts of Pd/C, Pt/C, Ru/C, Rh/C, CoMo/γ-Al2O3, Mo2C, MoS2, and activated carbon were screened. Mo2C was identified as the most suitable catalyst. With Mo2C catalyst, influence of reaction conditions on the yield and properties of the hydropyrolysis oil (HPO) was examined. Temperature was the most influential factor affecting the yield and quality of the HPO. Higher temperature will produce HPO with higher C and H content and lower N and O content but at the cost of lowering the yield of HPO. Mo2C promoted the in situ deoxygenation and desulfurization of the HPO which has a HHVs varying between 35.3 and 39.3 MJ/kg. The highest energy recovery of 87.5% was achieved. Thus, this work shows that the catalytic hydropyrolysis is an effective way to produce high quality bio-oil from microalgae.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Kaliumnitrat, ACS reagent, ≥99.0%
Sigma-Aldrich
Kaliumnitrat, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99%
Sigma-Aldrich
Kaliumnitrat, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Kaliumnitrat, BioReagent, suitable for cell culture, suitable for plant cell culture
Sigma-Aldrich
Kaliumnitrat, 99.999% trace metals basis
Sigma-Aldrich
Kaliumnitrat, BioXtra, ≥99.0%
Sigma-Aldrich
Kaliumnitrat, 99.99% trace metals basis
Supelco
Mettler-Toledo-Kalibriersubstanz ME 51143095, Kaliumnitrat, traceable to primary standards (LGC)