Merck
  • ABCG2 regulates the pattern of self-renewing divisions in cisplatin-resistant non-small cell lung cancer cell lines.
ABCG2 regulates the pattern of self-renewing divisions in cisplatin-resistant non-small cell lung cancer cell lines.
Oncology reports (2014-09-10)
Ying Tang, Jingming Hou, Guanghui Li, Zongchang Song, Xiaojing Li, Cui Yang, Wenying Liu, Yide Hu, Yu Xu
ZUSAMMENFASSUNG
Overexpression of ABCG2 is considered a major mechanism of cancer drug resistance. Recent studies have shown that ABCG2 can regulate the switch between symmetric and asymmetric cell division in adult stem cells; however, the relationship between ABCG2 and cell division in drug-resistant cancer cells remains to be determined. In the present study, we demonstrated that ABCG2 is involved in the cell division of drug-resistant cancer cells. We first established drug-resistant H460 and A549 cell lines by repeated exposure to cisplatin and found that the expression of ABCG2 in these cell lines was significantly increased. As evidenced by PKH-26 staining, these drug-resistant cell lines favored symmetric division, which differed from the asymmetric division of the parental cells. Furthermore, we established stable ABCG2‑overexpressing and stable shRNA-ABCG2‑knockdown cell lines to evaluate the potential role of ABCG2 in cancer cell division. The results showed that overexpression of ABCG2 in A549 parental cells significantly increased the proportion of symmetric division, whereas knockdown of ABCG2 in drug-resistant A549 cells significantly increased the proportion of asymmetric division. Taken together, our findings suggest that ABCG2 is involved in the modulation of cancer drug resistance by regulating the pattern of cell division. The present study provides novel insight into the role of ABCG2 in cancer treatment resistance.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Propidiumjodid, ≥94.0% (HPLC)
Sigma-Aldrich
cis-Diamminplatin(II)-dichlorid, crystalline
Sigma-Aldrich
Propidiumjodid -Lösung
Sigma-Aldrich
Propidiumjodid, ≥94% (HPLC)
Cisplatin, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
cis-Diaminplatin(II)-dichlorid, ≥99.9% trace metals basis
Sigma-Aldrich
trans-Platin(II)diammindichlorid
Cisplatin Unreinheit A, European Pharmacopoeia (EP) Reference Standard
USP
Transplatin, United States Pharmacopeia (USP) Reference Standard