Merck
  • N-Phenethyl caffeamide and photodamage: protecting skin by inhibiting type I procollagen degradation and stimulating collagen synthesis.
N-Phenethyl caffeamide and photodamage: protecting skin by inhibiting type I procollagen degradation and stimulating collagen synthesis.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association (2014-07-16)
Hsiu-Mei Chiang, Chien-Wen Chen, Tzu-Yu Lin, Yueh-Hsiung Kuo
ZUSAMMENFASSUNG
Skin is mainly damaged by genetic and environmental factors such as ultraviolet (UV) light and pollutants. UV light is a well-known factor that causes various types of skin damage and premature aging. Reactive oxygen species (ROS) are commonly involved in the pathogenesis of skin damage by activating the metalloproteinases that break down type I collagen. This study investigated the antioxidant and antiphotodamage activity and mechanisms of N-phenethyl caffeamide (K36) in human skin fibroblasts. The results indicated that K36 demonstrated strong 1,1-diphenyl-2-picrylhydrazyl radical (DPPH) scavenging activity, which dose-dependently reduced the production of UVB-induced intracellular ROS in human dermal fibroblasts. K36 prevented UVB-irradiation-induced type I collagen degradation by inhibiting the expression of matrix metalloproteins-1, -3, and -9 and the phosphorylation of mitogen-activated protein (MAP) kinases. Furthermore, K36 elevated collagen synthesis in skin fibroblasts by inhibiting UVB-induced Smad7 overexpression. K36 downregulated the expression of the transcription factor, activator protein-1 (AP-1). Our results indicated that K36 exhibited antioxidant properties and prevented skin collagen degradation caused by UV exposure and the stimulation of collagen synthesis, which suggests the potential use of K36 in preventing photodamage.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
2′,7′-Dichlorfluorescein-Diacetat, ≥97%
Sigma-Aldrich
Phenylmethansulfonylfluorid, ≥98.5% (GC)
Sigma-Aldrich
Propylenglycol, ≥99.5% (GC), FCC, FG
Sigma-Aldrich
Propylenglykol, meets USP testing specifications
Supelco
Propylenglykol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Phenylmethansulfonylfluorid, ≥99.0% (T)
Sigma-Aldrich
1,2-Propandiol, ACS reagent, ≥99.5%
USP
Propylenglycol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
1,2-Propandiol, ReagentPlus®, 99%
Sigma-Aldrich
1,2-Propandiol, puriss. p.a., ACS reagent, ≥99.5% (GC)
Sigma-Aldrich
1,2-Propandiol, meets analytical specification of Ph. Eur., BP, USP, ≥99.5%
Supelco
1,2-Propandiol, analytical standard
Propylenglycol, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
1,2-Propandiol, tested according to Ph. Eur.