Merck
  • Thioredoxin-interacting protein mediates NALP3 inflammasome activation in podocytes during diabetic nephropathy.
Thioredoxin-interacting protein mediates NALP3 inflammasome activation in podocytes during diabetic nephropathy.
Biochimica et biophysica acta (2014-07-16)
Pan Gao, Xian-Fang Meng, Hua Su, Fang-Fang He, Shan Chen, Hui Tang, Xiu-Juan Tian, Di Fan, Yu-Mei Wang, Jian-She Liu, Zhong-Hua Zhu, Chun Zhang
ZUSAMMENFASSUNG
Numerous studies have shown that the NALP3 inflammasome plays an important role in various immune and inflammatory diseases. However, whether the NALP3 inflammasome is involved in the pathogenesis of diabetic nephropathy (DN) is unclear. In our study, we confirmed that high glucose (HG) concentrations induced NALP3 inflammasome activation both in vivo and in vitro. Blocking NALP3 inflammasome activation by NALP3/ASC shRNA and caspase-1 inhibition prevented IL-1β production and eventually attenuated podocyte and glomerular injury under HG conditions. We also found that thioredoxin (TRX)-interacting protein (TXNIP), which is a pro-oxidative stress and pro-inflammatory factor, activated NALP3 inflammasome by interacting with NALP3 in HG-exposed podocytes. Knocking down TXNIP impeded NALP3 inflammasome activation and alleviated podocyte injury caused by HG. In summary, the NALP3 inflammasome mediates podocyte and glomerular injury in DN, moreover, TXNIP participates in the formation and activation of the NALP3 inflammasome in podocytes during DN, which represents a novel mechanism of podocyte and glomerular injury under diabetic conditions.