Merck
  • Protective effects of ectoine on behavioral, physiological and biochemical parameters of Daphnia magna subjected to hydrogen peroxide.
Protective effects of ectoine on behavioral, physiological and biochemical parameters of Daphnia magna subjected to hydrogen peroxide.
Comparative biochemistry and physiology. Toxicology & pharmacology : CBP (2015-02-24)
Adam Bownik, Zofia Stępniewska
ZUSAMMENFASSUNG
Ectoine (ECT) is an osmoprotectant produced by halophilic microorganisms inducing protective effects against various stressful factors. However, little is known about its influence on aquatic invertebrates subjected to hydrogen peroxide (H2O2)-a commonly used oxidative disinfectant. Therefore, the aim of our study was to determine the effects of H2O2 alone (at 5 and 10 mg/L) and in the combination with various concentrations of ECT (5, 10 and 25 mg/L) on behavioral, physiological and biochemical parameters of Daphnia magna. The following endpoints were determined: mortality, heart rate, thoracic limb movement, total glutathione (GSH)/oxidized glutathione (GSSG) ratio, catalase (CAT) activity and nitric oxide (NOx) level. The study showed that daphnids exposed to the combination of H2O2+ECT showed decreased mortality, attenuated inhibition of heart rate and thoracic limb activity, less decreased GSH/GSSG ratio, lower stimulation of CAT activity and NOx level when compared to the crustaceans exposed to H2O2 alone. The most pronounced alleviation of toxic effects was observed in the combination of 5 mg/L H2O2+25 mg/L ECT. The results suggest that protective effects of ECT in D. magna subjected to H2O2 may be related to antioxidative properties of the osmoprotectant.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Wasserstoffperoxid -Lösung, contains inhibitor, 30 wt. % in H2O, ACS reagent
Sigma-Aldrich
Wasserstoffperoxid -Lösung, 30 % (w/w) in H2O, contains stabilizer
Sigma-Aldrich
Ammoniummolybdat Tetrahydrat, ACS reagent, 81.0-83.0% MoO3 basis
Sigma-Aldrich
L-Glutathion reduziert, suitable for cell culture, BioReagent, ≥98.0%, powder
Sigma-Aldrich
L-Glutathion reduziert, ≥98.0%
Sigma-Aldrich
Wasserstoffperoxid -Lösung, 50 wt. % in H2O, stabilized
Sigma-Aldrich
Ammoniummolybdat Tetrahydrat, ACS reagent, 99.98% trace metals basis
Sigma-Aldrich
Ammoniummolybdat Tetrahydrat, BioUltra, ≥99.0% (T)
Sigma-Aldrich
Ammoniummolybdat, 99.98% trace metals basis
Millipore
Wasserstoffperoxid -Lösung, 3%, suitable for microbiology
Supelco
Wasserstoffperoxid -Lösung, ≥30%, for trace analysis
Sigma-Aldrich
Wasserstoffperoxid -Lösung, contains inhibitor, 30 wt. % in H2O, meets USP testing specifications
Sigma-Aldrich
Ammoniummolybdat Tetrahydrat, BioReagent, suitable for cell culture, suitable for insect cell culture, 81.0-83.0% MoO3 basis
Supelco
Glutathion, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Wasserstoffperoxid -Lösung, 34.5-36.5%
Supelco
Wasserstoffperoxid -Lösung, 30 % (w/w), for ultratrace analysis
Sigma-Aldrich
L-Glutathion reduziert, BioXtra, ≥98.0%
Sigma-Aldrich
Ammoniummolybdat, meets USP testing specifications
Sigma-Aldrich
Ectoin, ≥95.0% (HPLC)
Sigma-Aldrich
L-Glutathion, oxidiert Dinatriumsalz, ≥98%, powder
Sigma-Aldrich
Ectoin, osmoprotectant
Glutathion, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
L-Glutathion, oxidiert Dinatriumsalz, BioReagent, suitable for cell culture
Sigma-Aldrich
Wasserstoffperoxid -Lösung, tested according to Ph. Eur.