Merck
  • Sustained reprogramming of the estrogen response after chronic exposure to endocrine disruptors.
Sustained reprogramming of the estrogen response after chronic exposure to endocrine disruptors.
Molecular endocrinology (Baltimore, Md.) (2015-01-17)
Andrea R Patterson, Xiaokui Mo, Ali Shapiro, Karen E Wernke, Trevor K Archer, Craig J Burd
ZUSAMMENFASSUNG
The pervasive nature of estrogenic industrial and dietary compounds is a growing health concern linked to cancer, obesity, and neurological disorders. Prior analyses of endocrine disruptor action have focused primarily on the short-term consequences of exposure. However, these studies are unlikely to reflect the consequences of constant exposures common to industrialized countries. Here we examined the global effects of long-term endocrine disruption on gene transcription and estrogen signaling. Estrogen-dependent breast cancer cell lines were chronically treated with physiologically relevant levels of bisphenol A or genistein for more than 70 passages. Microarray analysis demonstrated global reprogramming of the transcriptome when compared with a similarly cultured control cell line. Estrogen-responsive targets showed diminished expression in both the presence and absence of estrogen. Estrogen receptor recruitment, H3K4 monomethylation, and deoxyribonuclease accessibility were reduced at nearby response elements. Based on these observations, we investigated the potential of long-term endocrine disruptor exposure to initiate persistent transcriptional reprogramming. Culture of chronically exposed cell lines in the absence of the endocrine disruptors did not reverse many of the signaling defects that accumulated during treatment. Taken together, these data demonstrate that chronic exposure to endocrine disrupting compounds can permanently alter physiological hormone signaling.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Ethyl alcohol, Pure 200 proof, Molecular Biology
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Sigma-Aldrich
1,2-Dichlorethan, ACS reagent, ≥99.0%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure 190 proof, for molecular biology
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
1,2-Dichlorethan, anhydrous, 99.8%
Sigma-Aldrich
Ethanol, BioUltra, Molecular Biology, ≥99.8%, (absolute alcohol, without additive, A15 o1)
Supelco
Ethanol, wasserfrei, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ethanol, purum, fine spirit, denaturated with 4.8% methanol, F25 METHYL1, ~96% (based on denaturant-free substance)
Supelco
Ethanol, standard for GC
Sigma-Aldrich
Phenolrot, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Phenolrot, ACS reagent
Sigma-Aldrich
Ethanol
Supelco
Ethanol -Lösung, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
Ethanol, tested according to Ph. Eur.
Supelco
1,2-Dichlorethan, analytical standard
USP
Dehydrated Alcohol, United States Pharmacopeia (USP) Reference Standard