Merck
Pistachio (Pistacia vera L.) gum: a potent inhibitor of reactive oxygen species.
Journal of enzyme inhibition and medicinal chemistry (2014-06-19)
M Hilal Sehitoglu, Hatice Han, Pınar Kalin, İlhami Gülçin, Ali Ozkan, Hassan Y Aboul-Enein
ZUSAMMENFASSUNG
In the present study, in order to evaluate antioxidant and radical scavenging properties of Pistachio gum (P-Gum), different bioanalytical methods such as DPPH(•) scavenging activity, DMPD(•+) radical scavenging activity, total antioxidant activity determination by ferric thiocyanate, reducing ability Fe(3+)-Fe(2+) transformation, Cuprac and FRAP assays, O2(•-) scavenging by riboflavin-methionine-illuminate system and ferrous ions (Fe(2+)) chelating activities by 2,2'-bipyridyl reagent were performed separately. P-Gum inhibited 54.2% linoleic acid peroxidation at 10 µg/ml concentration. On the other hand, BHA, BHT, α-tocopherol and trolox, pure antioxidant compounds, indicated inhibition of 80.3%, 73.5%, 36.2% and 72.0% on peroxidation of linoleic acid emulsion at the same concentration, respectively. In addition, all of sample had an effective DPPH(•), DMPD(•+) and O2(•-) scavenging, Fe(3+) reducing power by Fe(3+)-Fe(2+) transformation and FRAP assay, Cu(2+) reducing ability by Cuprac method and Fe(2+) chelating activities.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
2,6-Di-tert-butyl-4-Methylphenol, ≥99.0% (GC), powder
Sigma-Aldrich
α-Linoleic acid, ≥98%
Supelco
Butyliertes Hydroxytoluol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Butyliertes Hydroxytoluol, ≥99%, FCC, FG
Supelco
2,4,6-Tri-(2-pyridyl)-s-triazin, ≥98%
Sigma-Aldrich
(−)-Riboflavin, from Eremothecium ashbyii, ≥98%
Sigma-Aldrich
Linolsäure, liquid, BioReagent, suitable for cell culture
Sigma-Aldrich
Guajacol, oxidation indicator
Sigma-Aldrich
(−)-Riboflavin, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥98%
Supelco
Riboflavin (B2), analytical standard
Sigma-Aldrich
Guajacol, natural, ≥99%, FG
Supelco
(−)-Riboflavin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
N,N-Dimethyl-p-phenylendiamin, 97%
Millipore
2,3,5-Triphenyl-tetrazoliumchlorid -Lösung, suitable for microbiology, Filter sterilized solution that is recommended for the detection of microbial growth based on reduction of TTC
Supelco
2,4,6-Tri-(2-pyridyl)-s-triazin, for spectrophotometric det. of Fe, ≥99.0% (HPLC)
USP
Butyliertes Hydroxytoluol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
2,6-Di-tert-butyl-4-Methylphenol, purum, ≥99.0% (GC)
Sigma-Aldrich
DL-Methionin, ≥99%
Sigma-Aldrich
(−)-Riboflavin, meets USP testing specifications
Sigma-Aldrich
3-tert-Butyl-4-Hydroxyanisol, ≥98% (sum of isomers, GC), ≤10% 2-BHA basis (GC)
Sigma-Aldrich
Linolsäure, technical, 58-74% (GC)
Supelco
Linolsäure, analytical standard
USP
Riboflavin, United States Pharmacopeia (USP) Reference Standard
Supelco
Guajacol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
3-tert-Butyl-4-Hydroxyanisol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
DL-Methionin, 99%, FCC, FG
Riboflavin, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
2,6-Di-tert-butyl-4-Methylphenol, tested according to Ph. Eur.
Methionin, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
DL-Methionin, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99%