Merck
Genetic basis of a cognitive complexity metric.
PloS one (2015-04-11)
Narelle K Hansell, Graeme S Halford, Glenda Andrews, David H K Shum, Sarah E Harris, Gail Davies, Sanja Franic, Andrea Christoforou, Brendan Zietsch, Jodie Painter, Sarah E Medland, Erik A Ehli, Gareth E Davies, Vidar M Steen, Astri J Lundervold, Ivar Reinvang, Grant W Montgomery, Thomas Espeseth, Hilleke E Hulshoff Pol, John M Starr, Nicholas G Martin, Stephanie Le Hellard, Dorret I Boomsma, Ian J Deary, Margaret J Wright
ZUSAMMENFASSUNG
Relational complexity (RC) is a metric reflecting capacity limitation in relational processing. It plays a crucial role in higher cognitive processes and is an endophenotype for several disorders. However, the genetic underpinnings of complex relational processing have not been investigated. Using the classical twin model, we estimated the heritability of RC and genetic overlap with intelligence (IQ), reasoning, and working memory in a twin and sibling sample aged 15-29 years (N = 787). Further, in an exploratory search for genetic loci contributing to RC, we examined associated genetic markers and genes in our Discovery sample and selected loci for replication in four independent samples (ALSPAC, LBC1936, NTR, NCNG), followed by meta-analysis (N>6500) at the single marker level. Twin modelling showed RC is highly heritable (67%), has considerable genetic overlap with IQ (59%), and is a major component of genetic covariation between reasoning and working memory (72%). At the molecular level, we found preliminary support for four single-marker loci (one in the gene DGKB), and at a gene-based level for the NPS gene, having influence on cognition. These results indicate that genetic sources influencing relational processing are a key component of the genetic architecture of broader cognitive abilities. Further, they suggest a genetic cascade, whereby genetic factors influencing capacity limitation in relational processing have a flow-on effect to more complex cognitive traits, including reasoning and working memory, and ultimately, IQ.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Perchlorsäure, ACS reagent, 70%
Sigma-Aldrich
Perchlorsäure, 70%, 99.999% trace metals basis
Sigma-Aldrich
Perchlorsäure, ACS reagent, 60%
Supelco
Perchlorsäure-Konzentrat, 0.01 M HClO4 in water (0.01N), eluent for IC