Merck
  • Influenza virus uses its neuraminidase protein to evade the recognition of two activating NK cell receptors.
Influenza virus uses its neuraminidase protein to evade the recognition of two activating NK cell receptors.
The Journal of infectious diseases (2014-02-18)
Yotam Bar-On, Einat Seidel, Pinchas Tsukerman, Michal Mandelboim, Ofer Mandelboim
ZUSAMMENFASSUNG
Natural Killer (NK) cells play a central role in the defense against viral infections and in the elimination of transformed cells. The recognition of pathogen-infected and tumor cells is controlled by inhibitory and activating receptors. We have previously shown that among the activating (killer) NK cell receptors the natural cytotoxicity receptors, NKp44 and NKp46, interact with the viral hemagglutinin (HA) protein expressed on the cell surface of influenza-virus-infected cells. We further showed that the interaction between NKp44/NKp46 and viral HA is sialic-acid dependent and that the recognition of HA by NKp44 and NKp46 leads to the elimination of the infected cells. Here we demonstrate that the influenza virus developed a counter-attack mechanism and that the virus uses its neuraminidase (NA) protein to prevent the recognition of HA by both the NKp44 and NKp46 receptors, resulting in reduced elimination of the infected cells by NK cells.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Zanamivir, ≥98% (HPLC)
USP
Zanamivir, United States Pharmacopeia (USP) Reference Standard