Merck
  • Development of a prospective isopropyl alcohol-loaded pharmaceutical base using simultaneous in vitro/in vivo characterization methods of skin performance.
Development of a prospective isopropyl alcohol-loaded pharmaceutical base using simultaneous in vitro/in vivo characterization methods of skin performance.
Drug development and industrial pharmacy (2013-05-22)
Ivana Pantelic, Milica Lukic, Bojan Markovic, Lusiana, Christine Hoffmann, Christel Müller-Goymann, Jela Milic, Rolf Daniels, Snezana Savic
ZUSAMMENFASSUNG
Approaching of pharmaceutical and cosmetic industries in some aspects inevitably influence formulation of topical pharmaceuticals, urging researchers to introduce novel excipients with proven benefits over traditional ones. In that context, alkyl polyglucosides (APG) emerge as prominent natural-origin emulsifiers with numerous favorable features (biodegradability, dermatological acceptability, desirable sensory properties). To evaluate APG-stabilized bases (alone and upon addition of isopropyl alcohol) and their impact on skin performance. A simultaneous in vitro/in vivo skin absorption study was conducted to evaluate whether the tape stripping technique could be recommended as an in vivo tool for skin penetration assessment during formulation optimization process. After a comprehensive physicochemical characterization, biopharmaceutical properties of APG-bases versus reference ones were assessed through a combined in vitro (release/permeation) and in vivo approach. Physicochemical characterization revealed substantial difference in structural ordering due to the formation of various mesomorphic phases. The enhancer-loaded APG base resulted in significantly higher drug levels at all depths into the stratum corneum, indicating that the selected enhancer along with specific colloidal structure has increased the extent of drug delivery. Results recommend the investigated emulsifier for stabilization of topical drug delivery systems, not only for their ability to sustain the addition of isopropyl alcohol which proved to be a valuable enhancer, but also satisfactory skin absorption and tolerability when compared to samples stabilized by conventional emulsifier. Tape stripping proved to be a useful and yet inexpensive tool for in vivo trials, able to discriminate subtle differences in dermal availability.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Glycerin, ACS reagent, ≥99.5%
Sigma-Aldrich
Glycerin, Molecular Biology, ≥99.0%
Sigma-Aldrich
Glycerin, ReagentPlus®, ≥99.0% (GC)
Sigma-Aldrich
Glycerin, ≥99.5%
Sigma-Aldrich
Glycerin -Lösung, 83.5-89.5% (T)
Sigma-Aldrich
Glycerin, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Sigma-Aldrich
Glycerin, BioUltra, Molecular Biology, anhydrous, ≥99.5% (GC)
Supelco
Glycerin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Glycerin, BioXtra, ≥99% (GC)
USP
Glycerin, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Glycerin, FCC, FG
Sigma-Aldrich
Glycerin, meets USP testing specifications
Supelco
Glycerin, analytical standard
Sigma-Aldrich
Glycerin, tested according to Ph. Eur., anhydrous