Merck
The effect of TSPP-mediated photodynamic therapy and Parecoxib in experimental tumours.
Life sciences (2014-10-07)
Tiberiu Popescu, Iuliana Nenu, Mihaela D Aldea, Diana Olteanu, Dan Gheban, Corina Tatomir, Pompei Bolfa
ZUSAMMENFASSUNG
The study investigated the effects of the combined treatment Parecoxib (Pcox) and 5,10,15,20-tetra-sulphonato-phenyl-porphyrin(TSPP)-mediated photodynamic therapy on Walker 256 carcinosarcoma. Five groups of male Wistar rats were used: the control group, treated with TSPP, group 2, irradiated 24 h thereafter, group 3, treated with Pcox and irradiated 24 h thereafter, groups 4 and 5 treated with combined therapies, TSPP and Pcox before irradiation, and Pcox 24 h after TSPP and irradiation respectively. Tumour inflammation, growth and non-growth factors, apoptosis/necrosis rate and oxidative/nitrosative stress markers were investigated. Malondialdehyde levels and cyclooxygenase (COX)-2 expression increased significantly in the group treated with Pcox after TSPP-PDT when compared with TSPP + IR group (p < 0.05, p < 0.001 respectively), in correlation with a decrease in glutathione levels (p < 0.05). The quantification of apoptosis, based on the TUNEL-assay, and necrosis rate revealed an increase of apoptotic/necrotic index in the same group (p < 0.05). On the other hand, Pcox administered before irradiation showed a significant increase in both vascular endothelial growth factor (VEGF) and COX-2 levels (p < 0.05) and in nitric oxide production (p < 0.01), when compared with the control group. The administration of Pcox after TSPP-mediated PDT showed promising antitumoural effects, leading to an increase in oxidative and nitrosative stress as well as apoptosis/necrosis rate in tumour tissue. These results show that combined regimens that involve selective COX-2 inhibitors administration after irradiation may improve the therapeutic effectiveness of PDT.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Ethyl alcohol, Pure 200 proof, Molecular Biology
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Sigma-Aldrich
1-Butanol, 99.9%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Guanidin -hydrochlorid, Molecular Biology, ≥99%
Sigma-Aldrich
Natriumdodecylsulfat, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
1-Butanol, ACS reagent, ≥99.4%
Sigma-Aldrich
Ethyl alcohol, Pure 190 proof, for molecular biology
Sigma-Aldrich
Natriumdodecylsulfat, ACS reagent, ≥99.0%
Sigma-Aldrich
Natriumdodecylsulfat -Lösung, BioUltra, Molecular Biology, 10% in H2O
Sigma-Aldrich
Trichloressigsäure, ACS reagent, ≥99.0%
Sigma-Aldrich
Phthaldialdehyd, ≥97% (HPLC), powder or crystals
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Guanidin -hydrochlorid, organic base and chaeotropic agent, ≥99% (titration)
Sigma-Aldrich
Natriumdodecylsulfat, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
1-Butanol, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Guanidin -hydrochlorid, ≥98%
Sigma-Aldrich
Trichloressigsäure -Lösung, 6.1 N
Sigma-Aldrich
Ethanol, BioUltra, Molecular Biology, ≥99.8%, (absolute alcohol, without additive, A15 o1)
Sigma-Aldrich
Natriumdodecylsulfat, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Natriumdodecylsulfat -Lösung, BioUltra, 20% in H2O
Sigma-Aldrich
2,4-Dinitrophenylhydrazin, reagent grade, 97%
Sigma-Aldrich
1-Butanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5% (GC)
Supelco
Natriumdodecylsulfat, dust-free pellets, suitable for electrophoresis, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
1-Butanol, anhydrous, 99.8%
Sigma-Aldrich
2-Thiobarbitursäure, ≥98%
Supelco
Natriumdodecylsulfat, suitable for ion pair chromatography, LiChropur, ≥99.0%
Supelco
Ethanol, wasserfrei, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Phthaldialdehyd, suitable for fluorescence, ≥99.0% (HPLC)