Merck
  • Comparative analysis of theophylline and cholera toxin in rat colon reveals an induction of sealing tight junction proteins.
Comparative analysis of theophylline and cholera toxin in rat colon reveals an induction of sealing tight junction proteins.
Pflugers Archiv : European journal of physiology (2014-02-12)
Alexander G Markov, Evgeny L Falchuk, Natalia M Kruglova, Oksana V Rybalchenko, Michael Fromm, Salah Amasheh
ZUSAMMENFASSUNG
Claudin tight junction proteins have been identified to primarily determine intestinal epithelial barrier properties. While functional contribution of single claudins has been characterized in detail, information on the interplay with secretory mechanisms in native intestinal epithelium is scarce. Therefore, effects of cholera toxin and theophylline on rat colon were analyzed, including detection of sealing claudins. Tissue specimens were stripped off submucosal tissue layers and mounted in Ussing chambers, and short-circuit current (ISC) and transepithelial resistance (TER) were recorded. In parallel, expression and localization of claudins was analyzed and histological studies were performed employing hematoxylin-eosin staining and light and electron microscopy. Theophylline induced a strong increase of ISC in colon tissue specimens. In parallel, a decrease of TER was observed. In contrast, cholera toxin did not induce a significant increase of ISC, whereas an increase of TER was detected after 120 min. Western blots of membrane fractions revealed an increase of claudin-3 and -4 after incubation with cholera toxin, and theophylline induced an increase of claudin-4. In accordance, confocal laser-scanning microscopy exhibited increased signals of claudin-3 and -4 after incubation with cholera toxin, and increased signals of claudin-4 after incubation with theophylline, within tight junction complexes. Morphological analyses revealed no general changes of tight junction complexes, but intercellular spaces were markedly widened after incubation with cholera toxin and theophylline. We conclude that cholera toxin and theophylline have different effects on sealing tight junction proteins in native colon preparations, which may synergistically contribute to transport functions, in vitro.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
DAPI, for nucleic acid staining