Merck
  • Tracheosyringeal nerve transection in juvenile male zebra finches decreases BDNF in HVC and RA and the projection between them.
Tracheosyringeal nerve transection in juvenile male zebra finches decreases BDNF in HVC and RA and the projection between them.
Neuroscience letters (2014-09-16)
Yu Ping Tang, Juli Wade
ZUSAMMENFASSUNG
This study investigated relationships among disruption of normal vocal learning, brain-derived neurotrophic factor (BDNF), and the morphology of song nuclei in juvenile male zebra finches. The tracheosyringeal nerves were bilaterally transected at post-hatching day 20-25, so that the animals could not properly develop species-typical vocalizations. BDNF protein and the projection from HVC to the robust nucleus of the arcopallium (RA) were quantified during the sensorimotor integration phase of song development. The manipulation decreased the number of BDNF cells in HVC and RA, the volume of these areas defined by BDNF labeling, and the projection from HVC to RA. BDNF was not affected in Area X or the lateral magnocellular nucleus of the anterior nidopallium (LMAN). Thus, inhibition of a bird's ability to practice and/or to hear its own typically developing song specifically diminishes BDNF expression in cortical motor regions required for song production.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Absolute - Acetone free
USP
Methylalkohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Supelco
Methanol, analytical standard
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Methanol -Lösung, suitable for NMR (reference standard), 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Supelco
4-tert.-Octylphenol-monoethoxylat -Lösung, 10 μg/mL in acetone, analytical standard