Merck
  • Time-dependent effects of L-tryptophan administration on urinary excretion of L-tryptophan metabolites.
Time-dependent effects of L-tryptophan administration on urinary excretion of L-tryptophan metabolites.
Journal of nutritional science and vitaminology (2014-10-10)
Chiaki Hiratsuka, Mitsue Sano, Tsutomu Fukuwatari, Katsumi Shibata
ZUSAMMENFASSUNG
We have previously reported that dietary supplementation with up to 5.0 g/d of L-tryptophan (L-Trp) for 21 d has no adverse effects, judging from the levels of general blood variables, in healthy women. We performed a randomized, double-blind, placebo-controlled, crossover intervention study in 17 apparently healthy Japanese women. The subjects were randomly assigned to receive a placebo (0 g/d) or 1.0, 2.0, 3.0, 4.0, or 5.0 g/d of L-Trp for 21 d each with a 5-wk washout period between trials. We examined the 24-h urine profiles on days -1 (1 d before starting L-Trp), 7, 14, and 21 to determine whether administration of L-Trp at doses of up to 5.0 g/d affects time-dependent urinary excretion of L-Trp or its metabolites in healthy women. The urinary excretion of L-Trp, kynurenic acid, 3-hydroxykynurenine, xanthurenic acid, 3-hydroxyanthranilic acid, quinolinic acid, N(1)-methylnicotinamide, N(1)-methyl-2-pyridone-5-carboxamide, and N(1)-methyl-4-pyridone-3-carboxamide increased in an L-Trp dose-dependent manner on day 7. The amount of urinary excretion of these compounds was unchanged on days 14 and 21. The urinary excretion of serotonin, 5-hydroxyindole-3-acetic acid, 2-oxoadipic acid, and nicotinamide was unaffected by L-Trp at any of the doses tested. L-Trp doses had weak effects on the urinary excretion of kynurenine and anthranilic acid. Based on these findings, we conclude that there are no time-dependent effects of L-Trp administration in urinary excretion of L-Trp metabolites. Additionally, L-Trp and its metabolites do not accumulate in the body.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Nicotinamid, BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Nicotinamid, ≥99.5% (HPLC)
Sigma-Aldrich
Kynurensäure, ≥98%
Sigma-Aldrich
(−)-Riboflavin, from Eremothecium ashbyii, ≥98%
Sigma-Aldrich
Kreatinin, anhydrous, ≥98%
Supelco
Niacinamid, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
(−)-Riboflavin, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥98%
Supelco
Riboflavin (B2), analytical standard
Sigma-Aldrich
Nicotinamid, ≥98% (HPLC), powder
Supelco
(−)-Riboflavin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Anthranilsäure, reagent grade, ≥98%
Sigma-Aldrich
5-Hydroxyindol-3-Essigsäure, ≥98% (HPLC), crystalline
Sigma-Aldrich
(−)-Riboflavin, meets USP testing specifications
USP
Niacinamid, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
2,3-Pyridindicarboxylsäure, 99%
Sigma-Aldrich
Niacinamid, meets USP testing specifications
USP
Riboflavin, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
3-Hydroxy-DL-Kynurenin
Sigma-Aldrich
Serotonin creatinine sulfate monohydrate, powder
Sigma-Aldrich
3-Hydroxyanthranilsäure, 97%
Supelco
Kreatinin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Anthranilsäure, puriss. p.a., ≥99.5% (T)
Riboflavin, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
2-Oxoadipinsäure, ≥95.0% (HPLC)
Sigma-Aldrich
Nicotinamid, ≥98.5% (HPLC)
Nicotinamid, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
DL-Kynurenin, ≥95.0% (NT)
Riboflavin für die Peakidentifizierung, European Pharmacopoeia (EP) Reference Standard