Merck
Plasma cathepsin L: a prognostic marker for pancreatic cancer.
World journal of gastroenterology (2014-12-18)
Nidhi Singh, Prasenjit Das, Surabhi Gupta, Vikas Sachdev, Siddhartha Srivasatava, Siddhartha Datta Gupta, Ravindra Mohan Pandey, Peush Sahni, Shyam S Chauhan, Anoop Saraya
ZUSAMMENFASSUNG
To assess the prognostic significance of cathepsin L, a cysteine protease that degrades the peri-tumoral tissue, in patients with pancreatic cancer. Plasma samples from 127 pancreatic cancer patients were analyzed for cathepsin L levels by ELISA. Out of these patients, 25 underwent surgery and their paraffin-embedded tissue was analyzed for cathepsin L expression by immunohistochemistry. Survival of patients and clinicopathological parameters was correlated with cathepsin L expression in plasma and tissue using appropriate statistical analysis. The mean (± SD) cathepsin L in plasma samples of pancreatic cancer patients was 5.98 ± 2.5 ng/mL that was significantly higher compared to the levels in healthy controls (3.83 ± 0.45) or chronic pancreatitis patients (3.97 ± 1.06). Using ROC curve, a cut-off level of 5.0 ng/mL was decided for survival analysis. Elevated plasma levels of cathepsin L were found to be associated with poor prognosis (P = 0.01) in multivariate analysis. The plasma levels of the protease decreased after surgery. Though no significant correlation was seen between plasma and tissue expression of this protease, a trend did emerge that high cathepsin L expression in tissue correlated with its high levels in plasma. Cathepsin L levels in plasma of pancreatic cancer patients may be used as a potential prognostic marker for the disease.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Aceton, ACS reagent, ≥99.5%
Sigma-Aldrich
Aceton, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Aceton, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%
Sigma-Aldrich
L-Lysin, ≥98% (TLC)
Sigma-Aldrich
o-Xylen, reagent grade, ≥98.0%
Sigma-Aldrich
Aceton, ACS reagent, ≥99.5%
Supelco
Aceton, analytical standard
Sigma-Aldrich
L-Lysin, crystallized, ≥98.0% (NT)
Sigma-Aldrich
Aceton, histological grade, ≥99.5%
Sigma-Aldrich
o-Xylen, anhydrous, 97%
Supelco
Aceton, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Aceton, United States Pharmacopeia (USP) Reference Standard
Supelco
o-Xylen, analytical standard
Supelco
L-Lysin, analytical standard
Sigma-Aldrich
Aceton, ≥99%, FCC, FG
Sigma-Aldrich
Aceton, natural, ≥97%
Sigma-Aldrich
Aceton, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Tris(tert-butoxy)silanol, 99.999%
Supelco
o-Xylen, Pharmaceutical Secondary Standard; Certified Reference Material