Merck
Inhibitory effects of juglanin on cellular senescence in human dermal fibroblasts.
Journal of natural medicines (2014-02-13)
Hyo Hyun Yang, Kyoung Hwangbo, Ming Shan Zheng, Jong-Keun Son, Hwa Young Kim, Suk Hwan Baek, Hyung Chul Choi, So Young Park, Jae-Ryong Kim
ZUSAMMENFASSUNG
Cellular senescence contributes to tissue and organismal aging, tumor suppression and progress, tissue repair and regeneration, and age-related diseases. Thus, aging intervention might be a promising target for treatment and prevention of diverse age-related diseases. In the present study, we investigated whether juglanin purified from the crude extract of Polygonum aviculare exerted inhibitory activity against cellular senescence in human dermal fibroblasts (HDFs). Juglanin decreased senescence-associated β-galactosidase activity (SA-β-gal) and the level of reactive oxygen species in senescent cells induced by adriamycin treatment. Juglanin also repressed SA-β-gal activity in HDFs under replicative senescence. These results suggest that juglanin represses cellular senescence in HDFs and might be useful for the development of dietary supplements or cosmetics that alleviate tissue aging or age-related diseases.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
N-Acetyl-L-Cystein, BioReagent, suitable for cell culture
Sigma-Aldrich
N-Acetyl-L-Cystein, Sigma Grade, ≥99% (TLC), powder
Sigma-Aldrich
Luminol, 97%
Sigma-Aldrich
Rapamycin, Ready Made Solution, 2.5 mg/mL in DMSO (2.74 mM), from Streptomyces hygroscopicus
Sigma-Aldrich
Luminol, ≥97% (HPLC)
USP
Acetylcystein, United States Pharmacopeia (USP) Reference Standard
Supelco
N-Acetyl-L-Cystein, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
N-Acetyl-L-Cystein, BioXtra, ≥99% (TLC)
Supelco
Rapamycin, VETRANAL®, analytical standard
Acetylcystein, European Pharmacopoeia (EP) Reference Standard